Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02262947_cn
          See other UHMK1 CNV Assays ›
          Gene Symbol
          UHMK1
          Assay Reference Genome
          Location

          Chr.1:162523321 on build GRCh38
          Cytoband
          Assay ID Hs02262947_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          127933

          Gene Name:

          U2AF homology motif kinase 1

          Gene Aliases:

          KIS, KIST, P-CIP2

          Location:

          Chr.1:162497174-162529631 on Build GRCh38

          Assay Gene Location:

          Within Exon 9
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          UHMK1 NM_001184763.1 8 1886 NP_001171692.1
          NM_144624.2 7 2138 NP_653225.2
          NM_175866.5 8 2214 NP_787062.1
          AJ536197.1 8 2176 CAD60192.1
          AL834136.1 7 2612 CAI46199.1
          BC014917.1 7 1716 AAH14917.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6332269 Chr.1:160388767 - 162811116 on Build GRCh38 Duplication LOC105371470 CD48 PFDN2 USP21 PCP4L1 C1orf226 USF1 TOMM40L RPL31P11 ITLN2 F11R FCGR3A FCER1G B4GALT3 KLHDC9 DUSP12 MIR5187 MIR4654 OLFML2B UHMK1 LOC100422526 DEDD ATF6-DT CD84-AS1 FCRLA ARHGAP30 ATF6 NDUFS2 LY9 ADAMTS4 VANGL2 SDHC FCGR2B NOS1AP FCRLB CD244 PPOX NECTIN4-AS1 HSPA6 FCGR2C FCGR2A HSD17B7 LOC101928372 NECTIN4 APOA2 HSPA7 SH2D1B CD84 DDR2 UFC1 MPZ FCGR3B SLAMF7 SLAMF6 LOC124904439 CFAP126 NR1I3 TSTD1 ITLN1 SPATA46 SLAMF1 MIR556 NIT1 LOC112543491 UAP1
          esv2758973 Chr.1:162094678 - 162533566 on Build GRCh38 Gain SH2D1B NOS1AP UHMK1 SPATA46 C1orf226 MIR556 MIR4654
          nsv516482 Chr.1:162138329 - 162905405 on Build GRCh38 Gain+Loss SH2D1B CCDC190 DDR2 NOS1AP UHMK1 SPATA46 C1orf226 MIR556 HSD17B7 UAP1 MIR4654

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-serine phosphorylation
          neuron projection development
          positive regulation of translational initiation
          regulation of protein export from nucleus
          regulation of cell cycle
          nucleotide binding
          nucleic acid binding
          RNA binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          ribonucleoprotein complex binding
          protein serine kinase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline