Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02269694_cn
          See other MARK3 CNV Assays ›
          Gene Symbol
          MARK3
          Assay Reference Genome
          Location

          Chr.14:103491935 on build GRCh38
          Cytoband
          Assay ID Hs02269694_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          4140

          Gene Name:

          microtubule affinity regulating kinase 3

          Gene Aliases:

          CTAK1, KP78, PAR1A, Par-1a, VIPB

          Location:

          Chr.14:103385415-103503831 on Build GRCh38

          Assay Gene Location:

          Within Exon 18
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MARK3 NM_001128918.3 15 2360 NP_001122390.2
          NM_001128919.3 15 2360 NP_001122391.2
          NM_001128920.3 15 2312 NP_001122392.2
          NM_001128921.3 14 2123 NP_001122393.2
          NM_001411055.1 14 2306 NP_001397984.1
          NM_001411056.1 14 2168 NP_001397985.1
          NM_001437351.1
          NM_001437366.1 14 2306
          NM_001437367.1 15 2312
          NM_001437368.1 14 2120
          NM_001437369.1 14 2120
          NM_001438023.1 14 2168
          NM_002376.7 15 2360 NP_002367.5
          XM_005267641.6 15 2360 XP_005267698.2
          XM_005267642.6 15 2312 XP_005267699.2
          XM_005267643.6 XP_005267700.2
          XM_011536769.4 14 1523 XP_011535071.1
          XM_017021291.3 15 2312 XP_016876780.1
          XM_017021292.3 15 2312 XP_016876781.1
          XM_017021294.3 14 2168 XP_016876783.1
          XM_017021295.3 14 2168 XP_016876784.1
          XM_017021297.3 14 2120 XP_016876786.1
          XM_017021298.3 14 2168 XP_016876787.1
          XM_017021299.3 14 2120 XP_016876788.1
          XM_017021301.3 13 2119 XP_016876790.1
          XM_047431377.1 15 1802 XP_047287333.1
          XM_047431378.1 14 2306 XP_047287334.1
          XM_047431379.1 15 1802 XP_047287335.1
          XM_047431381.1 15 1857 XP_047287337.1
          XM_047431382.1 14 2306 XP_047287338.1
          XM_047431383.1 15 1801 XP_047287339.1
          XM_047431384.1 14 2306 XP_047287340.1
          XM_047431385.1 15 1857 XP_047287341.1
          XM_047431386.1 14 1806 XP_047287342.1
          XM_047431387.1 15 1754 XP_047287343.1
          XM_047431388.1 14 2258 XP_047287344.1
          XM_047431389.1 14 1539 XP_047287345.1
          XM_047431390.1 14 2120 XP_047287346.1
          XM_047431391.1 XP_047287347.1
          XM_047431392.1 XP_047287348.1
          XM_047431393.1 11 1383 XP_047287349.1
          XM_047431394.1 11 1370 XP_047287350.1
          XM_047431395.1 10 1289 XP_047287351.1
          XM_047431396.1 10 1278 XP_047287352.1
          AB209542.1 1 2539 BAD92779.1
          AF170723.1 6 800 AAD51631.1
          AF387637.1 15 1763 AAK82367.1
          AF465413.1 15 2255 AAL69982.1
          AK296643.1 8 1000 BAG59243.1
          AK307448.1 15 1790
          BC024773.1 15 2234 AAH24773.1
          BX161395.1 14 2104 CAD61882.1
          BX248250.1 6 781 CAD62578.1
          DQ368336.1 4 506 ABD15137.1
          M80359.1 15 1868 AAA59991.1
          U64205.1 15 2120 AAC15093.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv832883 Chr.14:103461350 - 103654616 on Build GRCh38 Loss KLC1 MARK3 TRMT61A BAG5 COA8 CKB
          nsv1054574 Chr.14:103210111 - 105365114 on Build GRCh38 Loss CDCA4 KLC1 MARK3 KIF26A-DT EIF5-DT MIR12121 MIR203B VESTAR NUDT14 C14orf180 TRMT61A EIF5 COA8 LINC02298 PLD4 LOC105370689 ADSS1 CLBA1 MIR6765 SIVA1 TMEM179 CKB GPR132 ZBTB42 AKT1 MIR4710 BAG5 KIF26A PACS2 MIR203A ZFYVE21 AHNAK2 PPP1R13B-DT LINC02691 LINC02280 LOC105378183 CEP170B ATP5MJ SNORA28 JAG2 BTBD6 LOC124903408 TDRD9 ASPG INF2 BRF1 PPP1R13B XRCC3 RD3L
          nsv1044967 Chr.14:103488390 - 103540714 on Build GRCh38 Gain MARK3 TRMT61A CKB

          Back To Top

          More Information


          Additional Information:

          Set Membership:

          Intragenic Exonic DGV Variation Validated

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          protein phosphorylation
          regulation of G2/M transition of mitotic cell cycle
          negative regulation of hippo signaling
          intracellular signal transduction
          negative regulation of protein localization to nucleus
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          protein phosphatase binding
          tau protein binding
          tau-protein kinase activity
          protein serine kinase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline