Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02359019_cn
          See other UBE2N CNV Assays ›
          Gene Symbol
          UBE2N
          Assay Reference Genome
          Location

          Chr.12:93410888 on build GRCh38
          Cytoband
          Assay ID Hs02359019_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          7334

          Gene Name:

          ubiquitin conjugating enzyme E2 N

          Gene Aliases:

          HEL-S-71, UBC13, UBCHBEN, UBCHBEN; UBC13, UbcH-ben, UbcH13

          Location:

          Chr.12:93405684-93441947 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 2 - Exon 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          UBE2N NM_003348.4 NP_003339.1
          AK098233.1 1 411
          BC000396.2 AAH00396.1
          BC003365.1 AAH03365.1
          BC108704.1 AAI08705.1
          BT006873.1 AAP35519.1
          D83004.1 BAA11675.1
          FJ224354.1 ACI46046.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv983353 Chr.12:93409500 - 93411232 on Build GRCh38 Duplication UBE2N
          nsv1044304 Chr.12:93219056 - 93419239 on Build GRCh38 Gain LOC643339 NUDT4 UBE2N
          dgv1654n223 Chr.12:93217272 - 93436984 on Build GRCh38 Duplication LOC643339 NUDT4 UBE2N

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          double-strand break repair via homologous recombination
          DNA double-strand break processing
          DNA repair
          regulation of DNA repair
          postreplication repair
          ubiquitin-dependent protein catabolic process
          protein monoubiquitination
          cellular response to DNA damage stimulus
          protein ubiquitination
          cellular response to nutrient levels
          TORC1 signaling
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of DNA repair
          T cell receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          protein K63-linked ubiquitination
          antiviral innate immune response
          positive regulation of protein K63-linked ubiquitination
          positive regulation of intracellular signal transduction
          negative regulation of TORC1 signaling
          positive regulation of double-strand break repair
          nucleotide binding
          RNA binding
          ubiquitin-protein transferase activity
          protein binding
          ATP binding
          transferase activity
          ubiquitin protein ligase binding
          ubiquitin binding
          ubiquitin conjugating enzyme activity
          ubiquitin-protein transferase activator activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline