Search
Search

Entrez Gene ID: | 9236 |
Gene Name: | cell cycle progression 1 |
Gene Aliases: |
CPR8 |
Location: |
Chr.15:55355239-55408359 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCPG1 | NM_001204450.2 | 9 | 3427 | NP_001191379.1 |
| NM_001204451.2 | 10 | 2278 | NP_001191380.1 | |
| NM_004748.6 | 8 | 6930 | NP_004739.3 | |
| NM_020739.5 | 8 | 6556 | NP_065790.2 | |
| AA740595.1 | 1 | 144 |
Entrez Gene ID: | 100533483 |
Gene Name: | DNAAF4-CCPG1 readthrough (NMD candidate) |
Gene Aliases: |
DYX1C1-CCPG1 |
Location: |
Chr.15:55355223-55498584 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAAF4-CCPG1 | NR_037923.1 | 16 | 4696 |
Entrez Gene ID: | 9488 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class B |
Gene Aliases: |
DEE80, EIEE80, GPI-MT-III, PIG-B |
Location: |
Chr.15:55319222-55355648 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGB | NM_004855.5 | 12 | 1627 | NP_004846.4 |
| XM_005254795.6 | 10 | 1373 | XP_005254852.1 | |
| XM_011522235.4 | 12 | 1582 | XP_011520537.1 | |
| XM_011522236.4 | 11 | 1522 | XP_011520538.1 | |
| XM_011522237.3 | 11 | 1428 | XP_011520539.1 | |
| XM_017022730.1 | 11 | 1333 | XP_016878219.1 | |
| XM_017022731.2 | 12 | 1458 | XP_016878220.1 | |
| XM_047433363.1 | 12 | 1589 | XP_047289319.1 | |
| XM_047433364.1 | 11 | 1534 | XP_047289320.1 | |
| AK093138.1 | 3 | 1655 | ||
| AK223330.1 | 12 | 1619 | BAD97050.1 | |
| AK293666.1 | 9 | 1530 | BAG57112.1 | |
| AK314706.1 | 12 | 1889 | BAG37253.1 | |
| BC017711.1 | 12 | 1851 | AAH17711.1 | |
| D42138.1 | 12 | 1642 | BAA07709.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761899 | Chr.15:53592144 - 55643724 on Build GRCh38 | Gain+Loss |
|
| nsv4242082 | Chr.15:55003981 - 55380858 on Build GRCh38 | Duplication |
|
| nsv6515550 | Chr.15:55306219 - 55474102 on Build GRCh38 | Duplication |
|
| nsv520799 | Chr.15:55353463 - 55468324 on Build GRCh38 | Loss |
|
| nsv1043789 | Chr.15:55108674 - 55800256 on Build GRCh38 | Gain |
|
| nsv569523 | Chr.15:55263404 - 55544506 on Build GRCh38 | Gain |
|
| esv3569598 | Chr.15:55270429 - 55410270 on Build GRCh38 | Gain |
|
| nsv4247248 | Chr.15:55307775 - 55493742 on Build GRCh38 | Duplication |
|
Set Membership: |
|