Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02661698_cn
          See other DHTKD1 CNV Assays ›
          Gene Symbol
          DHTKD1
          Assay Reference Genome
          Location

          Chr.10:12097714 on build GRCh38
          Cytoband
          Assay ID Hs02661698_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          55526

          Gene Name:

          dehydrogenase E1 and transketolase domain containing 1

          Gene Aliases:

          AAKAD, AMOXAD, CMT2Q, E1a, OADC-E1, OADH-E1

          Location:

          Chr.10:12068954-12123221 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 7 - Exon 8
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          DHTKD1 NM_018706.7 NP_061176.4
          AK309810.1
          BC002477.2 AAH02477.1
          BC007955.2 AAH07955.1
          CR749726.1 CAH18489.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1035894 Chr.10:12039222 - 12127501 on Build GRCh38 Gain DHTKD1 UPF2
          nsv6141674 Chr.10:12058346 - 12124178 on Build GRCh38 Duplication DHTKD1
          nsv831785 Chr.10:11928851 - 12124076 on Build GRCh38 Gain DHTKD1 UPF2
          dgv673n100 Chr.10:12049505 - 12137652 on Build GRCh38 Gain SEC61A2 DHTKD1 MIR548AK
          nsv4173580 Chr.10:12058308 - 12124170 on Build GRCh38 Duplication DHTKD1
          dgv37n21 Chr.10:12024028 - 12123248 on Build GRCh38 Gain DHTKD1 UPF2
          dgv674n100 Chr.10:12077012 - 12134998 on Build GRCh38 Gain SEC61A2 DHTKD1 MIR548AK
          esv3622354 Chr.10:12050805 - 12129371 on Build GRCh38 Gain DHTKD1
          nsv6452283 Chr.10:12095578 - 12121488 on Build GRCh38 Deletion DHTKD1
          nsv4188028 Chr.10:12094656 - 12215246 on Build GRCh38 Duplication NUDT5 SEC61A2 CDC123 DHTKD1 MIR548AK
          nsv1035709 Chr.10:12054410 - 12183299 on Build GRCh38 Gain NUDT5 SEC61A2 DHTKD1 MIR548AK
          esv3693047 Chr.10:12058736 - 12117648 on Build GRCh38 Gain DHTKD1
          nsv6442697 Chr.10:12058331 - 12124163 on Build GRCh38 Duplication DHTKD1
          nsv1046396 Chr.10:12059681 - 12109949 on Build GRCh38 Gain DHTKD1
          esv3891755 Chr.10:12080356 - 12104859 on Build GRCh38 Loss DHTKD1

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          generation of precursor metabolites and energy
          glycolytic process
          oxoglutarate dehydrogenase (succinyl-transferring) activity
          protein binding
          oxidoreductase activity
          oxidoreductase activity, acting on the aldehyde or oxo group of donors, disulfide as acceptor
          thiamine pyrophosphate binding
          2-oxoadipate dehydrogenase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline