Search
Search

Entrez Gene ID: | 91419 |
Gene Name: | ATP23 metallopeptidase and ATP synthase assembly factor homolog |
Gene Aliases: |
KUB3, XRCC6BP1 |
Location: |
Chr.12:57941567-57959148 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP23 | NM_001320408.2 | 6 | 1245 | NP_001307337.1 |
| NM_001320409.2 | 5 | 1159 | NP_001307338.1 | |
| NM_001320410.2 | 7 | 1376 | NP_001307339.1 | |
| NM_033276.4 | 6 | 1205 | NP_150592.1 | |
| XM_011538976.3 | 5 | 943 | XP_011537278.1 | |
| XM_017020206.2 | 6 | 1096 | XP_016875695.1 | |
| AF078164.2 | AAD31085.1 | |||
| BC012776.2 | AAH12776.3 | |||
| BC115381.1 | AAI15382.1 | |||
| BC115382.1 | AAI15383.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559046 | Chr.12:57768302 - 58084815 on Build GRCh38 | Gain |
|
| nsv6466759 | Chr.12:57924023 - 58187129 on Build GRCh38 | Deletion |
|
| nsv975500 | Chr.12:57957269 - 57968890 on Build GRCh38 | Duplication |
|
| nsv523853 | Chr.12:57956848 - 57992939 on Build GRCh38 | Gain |
|
Set Membership: |
|