Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03406253_cn
          See other PRKCE CNV Assays ›
          Gene Symbol
          PRKCE
          Assay Reference Genome
          Location

          Chr.2:45710637 on build GRCh38
          Cytoband
          Assay ID Hs03406253_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5581

          Gene Name:

          protein kinase C epsilon

          Gene Aliases:

          PKCE, nPKC-epsilon

          Location:

          Chr.2:45651279-46187990 on Build GRCh38

          Assay Gene Location:

          Within Intron 8
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PRKCE NM_005400.3 NP_005391.1
          XM_005264428.2 XP_005264485.1
          XM_005264431.5 XP_005264488.1
          XM_011532971.4 XP_011531273.1
          XM_011532975.4 XP_011531277.1
          XM_011532978.3 XP_011531280.1
          XM_011532980.4 XP_011531282.1
          XM_017004488.3 XP_016859977.1
          XM_017004490.3 XP_016859979.1
          XM_017004492.3 XP_016859981.1
          XM_047445094.1 XP_047301050.1
          XM_047445095.1 XP_047301051.1
          XM_047445096.1 XP_047301052.1
          XM_047445097.1 XP_047301053.1
          XM_047445098.1 XP_047301054.1
          XM_047445099.1 XP_047301055.1
          XM_047445100.1 XP_047301056.1
          AK313842.1 BAG36574.1
          AL832345.1
          BC109033.2 AAI09034.1
          BC109034.2 AAI09035.1
          X65293.1 CAA46388.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv3852n223 Chr.2:45159446 - 45915141 on Build GRCh38 Duplication PRKCE SRBD1 LINC01121
          dgv3804n100 Chr.2:45180603 - 45750333 on Build GRCh38 Gain PRKCE SRBD1 LINC01121
          esv2751894 Chr.2:45141551 - 45772680 on Build GRCh38 Gain PRKCE SRBD1 LINC01121
          dgv1119e212 Chr.2:45181796 - 45749281 on Build GRCh38 Gain PRKCE SRBD1 LINC01121
          nsv1013787 Chr.2:45541783 - 45738705 on Build GRCh38 Gain PRKCE SRBD1
          nsv1008226 Chr.2:45706491 - 45786214 on Build GRCh38 Loss PRKCE
          nsv998852 Chr.2:45419685 - 46135538 on Build GRCh38 Gain PRKCE SRBD1
          esv3892048 Chr.2:45674616 - 45719660 on Build GRCh38 Loss PRKCE
          nsv4071903 Chr.2:45413666 - 46137516 on Build GRCh38 Duplication PRKCE SRBD1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          macrophage activation involved in immune response
          immune system process
          apoptotic process
          cell adhesion
          signal transduction
          toxin catabolic process
          positive regulation of epithelial cell migration
          positive regulation of fibroblast migration
          positive regulation of cell-substrate adhesion
          regulation of metal ion transport
          peptidyl-serine phosphorylation
          regulation of lipid metabolic process
          insulin secretion
          positive regulation of actin filament polymerization
          negative regulation of protein ubiquitination
          cell-substrate adhesion
          lipopolysaccharide-mediated signaling pathway
          positive regulation of insulin secretion
          positive regulation of synaptic transmission, GABAergic
          positive regulation of cytokinesis
          positive regulation of superoxide anion generation
          intracellular signal transduction
          locomotory exploration behavior
          TRAM-dependent toll-like receptor 4 signaling pathway
          Fc-gamma receptor signaling pathway involved in phagocytosis
          xenobiotic catabolic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to morphine
          positive regulation of MAPK cascade
          positive regulation of lipid catabolic process
          regulation of release of sequestered calcium ion into cytosol
          cell division
          establishment of localization in cell
          synaptic transmission, GABAergic
          regulation of insulin secretion involved in cellular response to glucose stimulus
          mucus secretion
          positive regulation of mucus secretion
          cellular response to ethanol
          cellular response to prostaglandin E stimulus
          cellular response to hypoxia
          positive regulation of wound healing
          negative regulation of sodium ion transmembrane transport
          positive regulation of protein localization to plasma membrane
          regulation of cation transmembrane transport
          nucleotide binding
          actin monomer binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein kinase C activity
          calcium-independent protein kinase C activity
          protein binding
          ATP binding
          enzyme activator activity
          zinc ion binding
          kinase activity
          transferase activity
          enzyme binding
          receptor activator activity
          ethanol binding
          metal ion binding
          14-3-3 protein binding
          protein serine kinase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline