Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03568422_cn
          See other MAPK9 CNV Assays ›
          Gene Symbol
          MAPK9
          Assay Reference Genome
          Location

          Chr.5:180250066 on build GRCh38
          Cytoband
          5q35.3
          Assay ID Hs03568422_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5601

          Gene Name:

          mitogen-activated protein kinase 9

          Gene Aliases:

          JNK-55, JNK2, JNK2A, JNK2ALPHA, JNK2B, JNK2BETA, PRKM9, SAPK, SAPK1a, p54a, p54aSAPK

          Location:

          Chr.5:180233594-180292071 on Build GRCh38

          Assay Gene Location:

          Within Intron 7
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MAPK9 NM_001135044.1 NP_001128516.1
          NM_001308244.1 NP_001295173.1
          NM_002752.4 NP_002743.3
          NM_139068.2 NP_620707.1
          NM_139069.2 NP_620708.1
          NM_139070.2 NP_620709.1
          XM_005265940.4 XP_005265997.1
          XM_006714891.2 XP_006714954.1
          XM_017009638.1 XP_016865127.1
          XM_017009639.1 XP_016865128.1
          XM_017009640.1 XP_016865129.1
          XM_017009641.1 XP_016865130.1
          XM_017009642.1 XP_016865131.1
          XM_017009643.1 XP_016865132.1
          AB451302.1
          AB451355.1
          AB451433.1
          AK289638.1
          BC032539.1 AAH32539.1
          CR536580.1 CAG38817.1
          EU927388.1
          HM036706.1
          HM036707.1
          HM036708.1
          HM036709.1
          L31951.1 AAA56831.1
          U09759.1 AAA74740.1
          U34821.1 AAC50606.1
          U35002.1 AAC50608.1
          U35003.1 AAC50609.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1033706 Chr.5:179975769 - 180379356 on Build GRCh38 Gain MAPK9 GFPT2 RASGEF1C MIR340 RNF130
          esv2759398 Chr.5:180107477 - 180302944 on Build GRCh38 Gain MAPK9 GFPT2 RASGEF1C

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          release of cytochrome c from mitochondria
          positive regulation of protein phosphorylation
          protein phosphorylation
          protein targeting to mitochondrion
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          response to stress
          JNK cascade
          JUN phosphorylation
          central nervous system development
          response to mechanical stimulus
          response to toxic substance
          positive regulation of gene expression
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of cell morphogenesis involved in differentiation
          response to amine
          neuron projection development
          positive regulation of prostaglandin biosynthetic process
          regulation of protein ubiquitination
          positive regulation of prostaglandin secretion
          positive regulation of chemokine production
          cellular response to UV
          Fc-epsilon receptor signaling pathway
          response to drug
          regulation of circadian rhythm
          positive regulation of nitric oxide biosynthetic process
          positive regulation of transcription, DNA-templated
          regulation of JNK cascade
          response to cadmium ion
          rhythmic process
          regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of nitric-oxide synthase biosynthetic process
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to growth factor stimulus
          positive regulation of podosome assembly
          positive regulation of apoptotic signaling pathway
          JUN kinase activity
          protein binding
          ATP binding
          transcription factor binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          mitogen-activated protein kinase kinase kinase binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline