Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03753447_cn
          See other PFKP CNV Assays ›
          Gene Symbol
          PFKP
          Assay Reference Genome
          Location

          Chr.10:3111469 on build GRCh38
          Cytoband
          Assay ID Hs03753447_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5214

          Gene Name:

          phosphofructokinase, platelet

          Gene Aliases:

          ATP-PFK, PFK-C, PFK-P, PFKF

          Location:

          Chr.10:3067548-3136805 on Build GRCh38

          Assay Gene Location:

          Within Intron 14
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PFKP NM_001242339.2 NP_001229268.1
          NM_001323067.2 NP_001309996.1
          NM_001323068.2 NP_001309997.1
          NM_001323069.2 NP_001309998.1
          NM_001323070.1 NP_001309999.1
          NM_001323071.2 NP_001310000.1
          NM_001323072.2 NP_001310001.1
          NM_001323073.1 NP_001310002.1
          NM_001323074.1 NP_001310003.1
          NM_001345944.1 NP_001332873.1
          NM_001410880.1 NP_001397809.1
          NM_002627.5 NP_002618.1
          XM_005252466.5 XP_005252523.1
          XM_006717449.2 XP_006717512.1
          XM_047425350.1 XP_047281306.1
          AK092597.1 BAG52577.1
          AK291841.1 BAF84530.1
          AK308003.1
          AK308226.1
          AK309692.1
          BC002536.1 AAH02536.1
          BC021698.1 AAH21698.1
          BC029138.1 AAH29138.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv549814 Chr.10:3055020 - 3170233 on Build GRCh38 Loss SNORD142 PITRM1-AS1 PFKP-DT PITRM1 PFKP LOC124902536
          esv3622165 Chr.10:3099237 - 3125437 on Build GRCh38 Gain PFKP
          dgv932n54 Chr.10:2382612 - 3253481 on Build GRCh38 Gain SNORD142 PITRM1-AS1 PFKP-DT LOC107983989 LINC02645 LINC02668 PITRM1 PFKP LOC124902536 LOC105376350 LOC105376353
          nsv428229 Chr.10:2618610 - 3156581 on Build GRCh38 Gain SNORD142 PITRM1-AS1 PFKP-DT PITRM1 PFKP LOC124902536 LOC105376350
          esv2730995 Chr.10:2770220 - 3706473 on Build GRCh38 Deletion SNORD142 PITRM1-AS1 PFKP-DT LINC02668 PITRM1 PFKP LOC124902536 LOC124902565 LOC105376360 LINC02669 LOC105376353
          nsv3247937 Chr.10:3092335 - 3114052 on Build GRCh38 Insertion PFKP
          esv3447223 Chr.10:3110560 - 3111458 on Build GRCh38 Duplication PFKP
          nsv6442021 Chr.10:3030896 - 3173781 on Build GRCh38 Duplication SNORD142 PITRM1-AS1 PFKP-DT PITRM1 PFKP LOC124902536
          esv2750891 Chr.10:3098894 - 3164173 on Build GRCh38 Loss SNORD142 PITRM1-AS1 PITRM1 PFKP
          nsv516811 Chr.10:3108122 - 3113598 on Build GRCh38 Loss PFKP
          nsv470913 Chr.10:3052993 - 3125271 on Build GRCh38 Loss PFKP-DT PFKP LOC124902536
          nsv6442520 Chr.10:2846098 - 3581354 on Build GRCh38 Duplication SNORD142 PITRM1-AS1 PFKP-DT LINC02668 PITRM1 PFKP LOC124902536 LOC124902565 LOC105376360 LINC02669 LOC105376353
          nsv549810 Chr.10:3035272 - 3691194 on Build GRCh38 Gain SNORD142 PITRM1-AS1 PFKP-DT LINC02668 PITRM1 PFKP LOC124902536 LOC124902565 LOC105376360 LINC02669 LOC105376353
          nsv6454219 Chr.10:2905224 - 3284256 on Build GRCh38 Duplication SNORD142 PITRM1-AS1 PFKP-DT LINC02668 PITRM1 PFKP LOC124902536 LOC105376353
          nsv6437272 Chr.10:3058401 - 3259500 on Build GRCh38 Deletion SNORD142 PITRM1-AS1 PFKP-DT LINC02668 PITRM1 PFKP LOC124902536 LOC105376353
          nsv549818 Chr.10:3081241 - 3227125 on Build GRCh38 Gain SNORD142 PITRM1-AS1 PITRM1 PFKP
          nsv4179484 Chr.10:2460844 - 3257750 on Build GRCh38 Duplication SNORD142 PITRM1-AS1 PFKP-DT LOC107983989 LINC02645 LINC02668 PITRM1 PFKP LOC124902536 LOC105376350 LOC105376353
          dgv935n54 Chr.10:3044294 - 3125271 on Build GRCh38 Loss PFKP-DT PFKP LOC124902536
          dgv38n206 Chr.10:2613782 - 3185857 on Build GRCh38 Duplication SNORD142 PITRM1-AS1 PFKP-DT PITRM1 PFKP LOC124902536 LOC105376350
          nsv6455282 Chr.10:3085901 - 3154200 on Build GRCh38 Deletion SNORD142 PITRM1-AS1 PITRM1 PFKP

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs79011666] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          fructose 6-phosphate metabolic process
          glycolytic process
          fructose 1,6-bisphosphate metabolic process
          canonical glycolysis
          cellular response to leukemia inhibitory factor
          nucleotide binding
          catalytic activity
          6-phosphofructokinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          identical protein binding
          macromolecular complex binding
          cadherin binding
          metal ion binding
          fructose-6-phosphate binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline