Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04199315_cn
          See other UHMK1 CNV Assays ›
          Gene Symbol
          UHMK1
          Assay Reference Genome
          Location

          Chr.1:162522069 on build GRCh38
          Cytoband
          Assay ID Hs04199315_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          127933

          Gene Name:

          U2AF homology motif kinase 1

          Gene Aliases:

          KIS, KIST, P-CIP2

          Location:

          Chr.1:162497174-162529631 on Build GRCh38

          Assay Gene Location:

          Within Intron 8
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          UHMK1 NM_001184763.1 NP_001171692.1
          NM_144624.2 NP_653225.2
          NM_175866.5 NP_787062.1
          AJ536197.1 CAD60192.1
          AK058195.1
          AK292369.1 BAF85058.1
          AL834136.1 CAI46199.1
          BC014917.1 AAH14917.1
          BC026046.1 AAH26046.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6332269 Chr.1:160388767 - 162811116 on Build GRCh38 Duplication CD244 RPL31P11 LOC105371470 ADAMTS4 APOA2 C1orf226 NDUFS2 ATF6 PFDN2 MPZ FCRLB HSPA7 NOS1AP PPOX LOC124904439 NECTIN4-AS1 USF1 LOC101928372 CD84-AS1 SLAMF1 CD48 LOC100422526 UAP1 FCGR2C HSPA6 SDHC PCP4L1 USP21 SH2D1B ARHGAP30 TOMM40L DUSP12 B4GALT3 DDR2 SPATA46 KLHDC9 FCGR3A MIR556 UHMK1 MIR4654 FCRLA CD84 TSTD1 FCGR2B FCGR2A DEDD FCGR3B UFC1 CFAP126 OLFML2B SLAMF6 F11R LOC112543491 ATF6-DT FCER1G VANGL2 MIR5187 NIT1 SLAMF7 NR1I3 ITLN1 HSD17B7 ITLN2 NECTIN4 LY9
          esv2758973 Chr.1:162094678 - 162533566 on Build GRCh38 Gain SH2D1B MIR556 SPATA46 UHMK1 NOS1AP MIR4654 C1orf226
          nsv516482 Chr.1:162138329 - 162905405 on Build GRCh38 Gain+Loss SH2D1B MIR556 DDR2 SPATA46 UHMK1 HSD17B7 CCDC190 NOS1AP UAP1 MIR4654 C1orf226

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs143566359,rs68036113] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-serine phosphorylation
          neuron projection development
          positive regulation of translational initiation
          regulation of protein export from nucleus
          regulation of cell cycle
          nucleotide binding
          nucleic acid binding
          RNA binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          ribonucleoprotein complex binding
          protein serine kinase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline