Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05562812_cn
          See other PPP6R2 CNV Assays ›
          Gene Symbol
          PPP6R2
          Assay Reference Genome
          Location

          Chr.22:50409196 on build GRCh38
          Cytoband
          22q13.33
          Assay ID Hs05562812_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          9701

          Gene Name:

          protein phosphatase 6 regulatory subunit 2

          Gene Aliases:

          KIAA0685, PP6R2, SAP190, SAPS2

          Location:

          Chr.22:50343317-50445090 on Build GRCh38

          Assay Gene Location:

          Within Intron 8
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PPP6R2 NM_001242898.1 NP_001229827.1
          NM_001242899.1 NP_001229828.1
          NM_001242900.1 NP_001229829.1
          NM_014678.4 NP_055493.2
          XM_006724431.2 XP_006724494.1
          XM_006724434.1 XP_006724497.1
          XM_006724435.2 XP_006724498.1
          XM_011530720.2 XP_011529022.1
          XM_011530721.2 XP_011529023.1
          XM_011530722.2 XP_011529024.1
          XM_011530723.2 XP_011529025.1
          XM_011530724.2 XP_011529026.1
          XM_011530726.2 XP_011529028.1
          XM_011530727.2 XP_011529029.1
          XM_011530728.2 XP_011529030.1
          XM_011530729.2 XP_011529031.1
          XM_011530730.2 XP_011529032.1
          XM_011530731.1 XP_011529033.1
          XM_011530732.2 XP_011529034.1
          XM_011530734.2 XP_011529036.1
          XM_011530735.2 XP_011529037.1
          XM_011530736.2 XP_011529038.1
          XM_011530737.2 XP_011529039.1
          XM_011530738.2 XP_011529040.1
          XM_011530739.2 XP_011529041.1
          XM_011530740.2 XP_011529042.1
          XM_017029116.1 XP_016884605.1
          XM_017029117.1 XP_016884606.1
          XM_017029118.1 XP_016884607.1
          XM_017029119.1 XP_016884608.1
          XM_017029120.1 XP_016884609.1
          XM_017029121.1 XP_016884610.1
          XM_017029122.1 XP_016884611.1
          XM_017029123.1 XP_016884612.1
          XM_017029124.1 XP_016884613.1
          XM_017029125.1 XP_016884614.1
          XM_017029126.1 XP_016884615.1
          XM_017029127.1 XP_016884616.1
          XM_017029128.1 XP_016884617.1
          XM_017029129.1 XP_016884618.1
          XM_017029130.1 XP_016884619.1
          XM_017029131.1 XP_016884620.1
          XM_017029132.1 XP_016884621.1
          XM_017029133.1 XP_016884622.1
          XM_017029134.1 XP_016884623.1
          XM_017029135.1 XP_016884624.1
          XM_017029136.1 XP_016884625.1
          XM_017029137.1 XP_016884626.1
          XM_017029138.1 XP_016884627.1
          AB014585.1
          AK302472.1
          BC000976.4 AAH00976.2
          BC006568.2 AAH06568.1
          BC032664.1 AAH32664.1
          BC041698.1 AAH41698.1
          BC052995.2 AAH52995.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv834238 Chr.22:50402690 - 50556965 on Build GRCh38 Loss SCO2 LMF2 NCAPH2 PPP6R2 ODF3B KLHDC7B LOC107985556 TYMP SBF1 MIOX ADM2 SYCE3
          nsv3668 Chr.22:50391793 - 50436883 on Build GRCh38 Deletion PPP6R2 LOC107985556

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          phosphatase modulator

          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline