Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05563804_cn
          See other PPP6R2 CNV Assays ›
          Gene Symbol
          PPP6R2
          Assay Reference Genome
          Location

          Chr.22:50346693 on build GRCh38
          Cytoband
          Assay ID Hs05563804_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          9701

          Gene Name:

          protein phosphatase 6 regulatory subunit 2

          Gene Aliases:

          KIAA0685, PP6R2, SAP190, SAPS2

          Location:

          Chr.22:50330774-50445090 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PPP6R2 NM_001242898.2 NP_001229827.1
          NM_001242899.2 NP_001229828.1
          NM_001242900.2 NP_001229829.1
          NM_001351641.2 NP_001338570.1
          NM_001351642.2 NP_001338571.1
          NM_001351643.2 NP_001338572.1
          NM_001351644.2 NP_001338573.1
          NM_001351645.2 NP_001338574.1
          NM_001351646.2 NP_001338575.1
          NM_001351647.2 NP_001338576.1
          NM_001351648.2 NP_001338577.1
          NM_001365836.1 NP_001352765.1
          NM_014678.5 NP_055493.2
          XM_006724431.4 XP_006724494.1
          XM_011530732.3 XP_011529034.1
          XM_011530734.3 XP_011529036.1
          XM_011530736.4 XP_011529038.1
          XM_011530737.3 XP_011529039.1
          XM_017029120.2 XP_016884609.1
          XM_047441642.1 XP_047297598.1
          XM_047441643.1 XP_047297599.1
          XM_047441644.1 XP_047297600.1
          XM_047441645.1 XP_047297601.1
          XM_047441646.1 XP_047297602.1
          XM_047441647.1 XP_047297603.1
          XM_047441648.1 XP_047297604.1
          XM_047441649.1 XP_047297605.1
          XM_047441650.1 XP_047297606.1
          XM_047441651.1 XP_047297607.1
          XM_047441652.1 XP_047297608.1
          XM_047441653.1 XP_047297609.1
          XM_047441654.1 XP_047297610.1
          XM_047441655.1 XP_047297611.1
          XM_047441656.1 XP_047297612.1
          XM_047441657.1 XP_047297613.1
          XM_047441658.1 XP_047297614.1
          XM_047441659.1 XP_047297615.1
          XM_047441660.1 XP_047297616.1
          XM_047441661.1 XP_047297617.1
          AB014585.1 BAA31660.3
          AK302472.1 BAH13719.1
          BC006568.2 AAH06568.1
          BC032664.1 AAH32664.1
          BC041698.1 AAH41698.1
          BC052995.2 AAH52995.1
          DB452735.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6554758 Chr.22:50311988 - 50385638 on Build GRCh38 Duplication PPP6R2 DENND6B
          esv26897 Chr.22:50343296 - 50348608 on Build GRCh38 Loss PPP6R2
          nsv955200 Chr.22:50337072 - 50346871 on Build GRCh38 Deletion PPP6R2
          nsv1073177 Chr.22:50342970 - 50346671 on Build GRCh38 Deletion PPP6R2
          nsv6548765 Chr.22:50310770 - 50396235 on Build GRCh38 Deletion PPP6R2 DENND6B
          nsv3244706 Chr.22:50343517 - 50360991 on Build GRCh38 Insertion PPP6R2
          nsv589221 Chr.22:50345877 - 50347289 on Build GRCh38 Loss PPP6R2
          nsv6535590 Chr.22:48991693 - 50757224 on Build GRCh38 Duplication ACR MLC1 TRABD-AS1 ARSA CHKB-CPT1B TRABD ALG12 MIR12114 CIMAP1B CHKB-DT TYMP CHKB NCAPH2 MAPK12 PPP6R2 SYCE3 MIOX KLHDC7B-DT MAPK8IP2 TUBGCP6 PANX2 LOC124905156 MOV10L1 MIR3667 MAPK11 TTLL8 CPT1B KLHDC7B PLXNB2 ADM2 ZBED4 RPL23AP82 LOC105377205 SCO2 LOC105373100 NHIP LOC105373091 CRELD2 IL17REL LMF2 SELENOO MIR6821 HDAC10 BRD1 DENND6B SBF1 PIM3 SHANK3 MIR3667HG LOC102724608
          dgv8138n54 Chr.22:50343738 - 50347021 on Build GRCh38 Gain+Loss PPP6R2

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of signal transduction
          protein binding
          protein phosphatase regulator activity
          protein phosphatase binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline