Search
Search

Entrez Gene ID: | 6128 |
Gene Name: | ribosomal protein L6 |
Gene Aliases: |
L6, SHUJUN-2, TAXREB107, TXREB1 |
Location: |
Chr.12:112405181-112418850 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL6 | NM_000970.4 | NP_000961.2 | ||
| NM_001024662.2 | NP_001019833.1 | |||
| NM_001320137.1 | NP_001307066.1 | |||
| NM_001320138.1 | NP_001307067.1 | |||
| NM_001320139.1 | NP_001307068.1 | |||
| NM_001320140.1 | NP_001307069.1 | |||
| NM_001320141.1 | NP_001307070.1 | |||
| NM_001320142.1 | NP_001307071.1 | |||
| XM_017019781.1 | XP_016875270.1 | |||
| XM_017019782.1 | XP_016875271.1 | |||
| AK298117.1 | ||||
| AK299470.1 | ||||
| AK311863.1 | ||||
| AY320410.1 | AAP73810.1 | |||
| BC022444.1 | AAH22444.1 | |||
| BC104824.1 | AAI04825.1 | |||
| BC104826.1 | AAI04827.1 | |||
| CD049618.1 | ||||
| D17554.1 | BAA04491.1 | |||
| S71022.1 | AAB30819.1 | |||
| X69391.1 | CAA49188.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv976652 | Chr.12:112404852 - 112406170 on Build GRCh38 | Duplication |
|
| esv3580466 | Chr.12:112403584 - 112419956 on Build GRCh38 | Loss |
|
Set Membership: |
|