Search
Search

Entrez Gene ID: | 55101 |
Gene Name: | ATP5S like |
Gene Aliases: |
- |
Location: |
Chr.19:41431318-41447822 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5SL | XM_011527065.2 | XP_011525367.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644380 | Chr.19:41443924 - 41501183 on Build GRCh38 | Loss |
|