Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07164523_cn
          See other ITCH CNV Assays ›
          Gene Symbol
          ITCH
          Assay Reference Genome
          Location

          Chr.20:34440788 on build GRCh38
          Cytoband
          20q11.22
          Assay ID Hs07164523_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          83737

          Gene Name:

          itchy E3 ubiquitin protein ligase

          Gene Aliases:

          ADMFD, AIF4, AIP4, NAPP1

          Location:

          Chr.20:34363235-34511773 on Build GRCh38

          Assay Gene Location:

          Within Intron 13
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          ITCH NM_001257137.2 NP_001244066.1
          NM_001257138.2 NP_001244067.1
          NM_001324197.1 NP_001311126.1
          NM_001324198.1 NP_001311127.1
          NM_031483.6 NP_113671.3
          XM_005260570.1 XP_005260627.1
          XM_011529079.1 XP_011527381.1
          XM_017028089.1 XP_016883578.1
          XM_017028090.1 XP_016883579.1
          XM_017028091.1 XP_016883580.1
          XM_017028092.1 XP_016883581.1
          AB056663.2 BAB39389.1
          AB209747.1 BAD92984.1
          AF038564.1 AAC04845.1
          AF095745.1 AAK39399.1
          AK297809.1
          AK304090.1
          AK315212.1
          BC011571.1 AAH11571.1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic non-DGV Variation

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          positive regulation of T cell anergy
          ubiquitin-dependent protein catabolic process
          apoptotic process
          inflammatory response
          Notch signaling pathway
          protein ubiquitination
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of type I interferon production
          protein K29-linked ubiquitination
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic process
          innate immune response
          positive regulation of protein catabolic process
          negative regulation of JNK cascade
          negative regulation of alpha-beta T cell proliferation
          viral entry into host cell
          negative regulation of defense response to virus
          defense response to virus
          nucleotide-binding oligomerization domain containing signaling pathway
          protein K63-linked ubiquitination
          protein K48-linked ubiquitination
          regulation of protein deubiquitination
          ubiquitin-protein transferase activity
          protein binding
          ligase activity
          ubiquitin-like protein transferase activity
          ribonucleoprotein complex binding
          CXCR chemokine receptor binding
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline