Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00646722_cn
          See other PRKCA CNV Assays ›
          Gene Symbol
          Prkca
          Assay Reference Genome
          Location

          Chr.11:107979158 on build GRCm38
          Cytoband
          11E1
          Assay ID Mm00646722_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          18750

          Gene Name:

          protein kinase C, alpha

          Gene Aliases:

          AI875142, Pkca

          Location:

          Chr.11:107933387-108343888 on Build GRCm38

          Assay Gene Location:

          Overlaps Intron 12 - Exon 12
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          Prkca NM_011101.3 NP_035231.2
          AK147084.1 BAE27664.1
          AK163950.1 BAE37546.1
          BC096493.1 AAH96493.1
          M25811.1 AAA39934.1
          X52684.1 CAA36907.1
          X52685.1 CAA36908.1

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          inactivation of MAPK activity
          response to reactive oxygen species
          angiogenesis
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          regulation of the force of heart contraction
          chondrocyte differentiation
          desmosome assembly
          protein phosphorylation
          negative regulation of protein kinase activity
          cellular calcium ion homeostasis
          apoptotic process
          regulation of muscle contraction
          cell adhesion
          learning or memory
          negative regulation of cell proliferation
          positive regulation of endothelial cell migration
          positive regulation of cardiac muscle hypertrophy
          phosphorylation
          negative regulation of translation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          central nervous system neuron axonogenesis
          positive regulation of cell migration
          neutrophil chemotaxis
          positive regulation of lipopolysaccharide-mediated signaling pathway
          response to estradiol
          negative regulation of glial cell apoptotic process
          histone H3-T6 phosphorylation
          intracellular signal transduction
          peptidyl-serine autophosphorylation
          positive regulation of blood vessel endothelial cell migration
          response to ethanol
          positive regulation of macrophage differentiation
          positive regulation of angiogenesis
          positive regulation of cell adhesion
          negative regulation of heart contraction
          positive regulation of exocytosis
          positive regulation of mitotic cell cycle
          negative regulation of glucose import
          negative regulation of insulin receptor signaling pathway
          protein autophosphorylation
          regulation of receptor-mediated endocytosis
          positive regulation of smooth muscle cell proliferation
          positive regulation of inflammatory response
          regulation of peptidyl-tyrosine phosphorylation
          induction of positive chemotaxis
          positive regulation of synapse assembly
          positive regulation of ERK1 and ERK2 cascade
          response to interleukin-1
          cellular response to carbohydrate stimulus
          regulation of platelet aggregation
          intrinsic apoptotic signaling pathway
          positive regulation of dense core granule biogenesis
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein kinase C activity
          calcium-dependent protein kinase C activity
          protein binding
          ATP binding
          zinc ion binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          enzyme binding
          histone kinase activity (H3-T6 specific)
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline