Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01283159_cn
          See other SAXO5 CNV Assays ›
          Gene Symbol
          SAXO5
          Assay Reference Genome
          Location

          Chr.19:7504214 on build GRCh38
          Cytoband
          Assay ID Hs01283159_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          374877

          Gene Name:

          stabilizer of axonemal microtubules 5

          Gene Aliases:

          C19orf45, TEX45

          Location:

          Chr.19:7497548-7508450 on Build GRCh38

          Assay Gene Location:

          Overlaps Exon 3 - Intron 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SAXO5 NM_198534.3 NP_940936.2
          AK093103.1 BAC04057.1
          BC029824.1 AAH29824.1
          BC034779.1 AAH34779.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv5524610 Chr.19:7503354 - 7504693 on Build GRCh38 Deletion SAXO5
          nsv828416 Chr.19:7483643 - 7519201 on Build GRCh38 Loss ZNF358 SAXO5 PEX11G
          nsv1060887 Chr.19:7089546 - 7861087 on Build GRCh38 Gain TRAPPC5 CLEC4GP1 CLEC4M RETN CLEC4G PCP2 FCER2 ARHGEF18-AS1 ARHGEF18 MIR6792 PET100 EVI5L ZNF358 SAXO5 XAB2 INSR MCEMP1 CAMSAP3 MCOLN1 PNPLA6 STXBP2 CD209 PEX11G
          nsv2400 Chr.19:7481597 - 7512937 on Build GRCh38 Insertion SAXO5 PEX11G
          esv3643587 Chr.19:7503335 - 7504677 on Build GRCh38 Loss SAXO5
          esv2751809 Chr.19:6896369 - 7816144 on Build GRCh38 Gain CLEC4GP1 CLEC4M PCP2 ZNF557 MBD3L4 MBD3L2B MBD3L3 ARHGEF18-AS1 ADGRE1 ARHGEF18 PET100 MBD3L2 ZNF358 XAB2 INSR MCEMP1 CAMSAP3 MCOLN1 CD209 MBD3L5 TRAPPC5 RETN CLEC4G FCER2 MARK2P21 MIR6792 SAXO5 PNPLA6 ADGRE4P STXBP2 PEX11G

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs138148015] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline