Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01697491_cn
          See other OR6F1 CNV Assays ›
          Gene Symbol
          OR6F1
          Assay Reference Genome
          Location

          Chr.1:247711971 on build GRCh38
          Cytoband
          Assay ID Hs01697491_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          343169

          Gene Name:

          olfactory receptor family 6 subfamily F member 1

          Gene Aliases:

          OR1-34, OR1-38, OST731

          Location:

          Chr.1:247711436-247716646 on Build GRCh38

          Assay Gene Location:

          Within Exon 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          OR6F1 NM_001005286.2 3 1227 NP_001005286.1
          BC137142.1 1 833 AAI37143.1
          BC137143.1 1 833 AAI37144.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv34503 Chr.1:247589905 - 248212768 on Build GRCh38 Gain OR1C1 OR2M5 OR14L1 OR13G1 OR14A2 OR14A16 OR2W3 LOC102724446 OR2L3 OR6F1 OR11L1 OR2M1P OR2L1P TRIM58 OR2L8 OR2M3 OR2T8 OR2L13 OR2AK2 OR2L2 OR9H1 OR2G3 OR14K1 OR2M2 OR2AJ1 OR2L5
          nsv5448739 Chr.1:247661000 - 248560600 on Build GRCh38 Duplication OR1C1 OR2M5 OR2T29 OR2T12 OR13G1 OR14A2 OR14A16 OR2W3 OR2T7 OR2L3 OR2T3 OR6F1 OR2T5 OR11L1 OR2M1P OR2M4 OR2T6 OR2T1 OR2L1P OR14C36 TRIM58 OR2L8 OR2T2 OR2M3 OR2M7 OR2G6 OR2T8 OR2T4 OR2L13 OR2AK2 OR2L2 OR9H1 OR14K1 OR2M2 OR2AJ1 OR2T33 OR2L5
          nsv6328482 Chr.1:247693201 - 247753300 on Build GRCh38 Duplication OR14A2 OR14K1 OR6F1
          esv2759014 Chr.1:247551283 - 248159014 on Build GRCh38 Gain+Loss OR1C1 OR2M5 OR14L1 OR13G1 OR14A2 OR14A16 OR2W3 LOC102724446 OR2L3 OR6F1 OR11L1 OR2M1P OR2L1P TRIM58 OR2L8 OR2T8 OR2G2 OR2L13 OR2AK2 OR2L2 OR9H1 OR2G3 OR14K1 OR2AJ1 GCSAML OR2L5
          nsv515858 Chr.1:247620588 - 248705477 on Build GRCh38 Gain+Loss OR1C1 OR2M5 OR2T10 OR14L1 OR2T29 OR2T27 OR2T12 OR13G1 OR14A2 OR14A16 OR2W3 OR2T7 LOC102724446 OR2L3 OR2T3 OR6F1 OR2T34 OR2T5 OR11L1 OR2M1P OR2M4 OR2T6 OR2T1 LYPD9P OR2L1P OR2T11 OR14I1 OR14C36 TRIM58 OR2L8 OR2T2 OR2M3 OR2M7 OR2G6 OR2T8 OR2T4 OR2L13 OR2AK2 OR2L2 OR9H1 OR2T35 OR14K1 OR2M2 OR2AJ1 OR2T33 OR2L5
          nsv4063384 Chr.1:247708498 - 247713598 on Build GRCh38 Duplication OR6F1
          esv3589522 Chr.1:247708531 - 247996788 on Build GRCh38 Gain OR11L1 OR2L1P OR1C1 TRIM58 OR2L8 OR2T8 OR14A2 OR14A16 OR2W3 OR2L13 OR2AK2 OR9H1 OR14K1 OR2AJ1 OR6F1
          nsv5445146 Chr.1:247660364 - 248129682 on Build GRCh38 Duplication OR11L1 OR2M1P OR2L1P OR1C1 TRIM58 OR2L8 OR2T8 OR13G1 OR14A2 OR14A16 OR2W3 OR2L13 OR2AK2 OR2L2 OR9H1 OR2L3 OR14K1 OR2AJ1 OR6F1 OR2L5
          nsv549597 Chr.1:247686171 - 248333561 on Build GRCh38 Gain OR1C1 OR2M5 OR2T12 OR14A2 OR14A16 OR2W3 OR2L3 OR6F1 OR11L1 OR2M1P OR2M4 OR2L1P TRIM58 OR2L8 OR2M3 OR2M7 OR2T8 OR2L13 OR2AK2 OR2L2 OR9H1 OR14K1 OR2M2 OR2AJ1 OR2T33 OR2L5
          dgv604n100 Chr.1:247654777 - 248129856 on Build GRCh38 Gain OR11L1 OR2M1P OR2L1P OR1C1 TRIM58 OR2L8 OR2T8 OR13G1 OR14A2 OR14A16 OR2W3 OR2L13 OR2AK2 OR2L2 OR9H1 OR2L3 OR14K1 OR2AJ1 OR6F1 OR2L5
          esv2762235 Chr.1:247654789 - 248129868 on Build GRCh38 Gain OR11L1 OR2M1P OR2L1P OR1C1 TRIM58 OR2L8 OR2T8 OR13G1 OR14A2 OR14A16 OR2W3 OR2L13 OR2AK2 OR2L2 OR9H1 OR2L3 OR14K1 OR2AJ1 OR6F1 OR2L5
          dgv605n100 Chr.1:247664654 - 248032454 on Build GRCh38 Gain OR11L1 OR2L1P OR1C1 TRIM58 OR2L8 OR2T8 OR13G1 OR14A2 OR14A16 OR2W3 OR2L13 OR2AK2 OR2L2 OR9H1 OR14K1 OR2AJ1 OR6F1 OR2L5
          nsv1013273 Chr.1:247656673 - 248784560 on Build GRCh38 Gain OR1C1 AHCYP8 OR2M5 OR2T10 OR2T29 OR2T27 OR2T12 OR13G1 OR14A2 OR14A16 OR2W3 OR2T7 OR2L3 OR2T3 OR6F1 OR2T34 OR2T5 OR11L1 OR2M1P OR2M4 OR2T6 OR2T1 LYPD9P OR2L1P OR2T11 OR14I1 OR14C36 TRIM58 OR2L8 OR2T2 OR2M3 OR2M7 OR2G6 OR2T8 LYPD8 OR2T4 OR2L13 OR2AK2 OR2L2 OR9H1 OR2T35 OR14K1 OR2M2 OR2AJ1 OR2T33 OR2L5
          dgv583n223 Chr.1:247688201 - 247888200 on Build GRCh38 Duplication OR11L1 OR1C1 TRIM58 OR9H1 OR14A2 OR14K1 OR6F1 OR14A16
          nsv4064677 Chr.1:247699306 - 248166944 on Build GRCh38 Duplication OR11L1 OR2M1P OR2L1P OR1C1 OR2M5 TRIM58 OR2L8 OR2T8 OR14A2 OR14A16 OR2W3 OR2L13 OR2AK2 OR2L2 OR9H1 OR2L3 OR14K1 OR2AJ1 OR6F1 OR2L5
          esv33129 Chr.1:246911128 - 248165533 on Build GRCh38 Gain+Loss OR2B11 MIR3916 AHCTF1 OR1C1 OR2M5 VN1R5 OR14L1 LINC02897 OR13G1 OR14A2 OR14A16 OR2W3 ZNF670-ZNF695 LOC102724446 FLJ39095 ZNF496-DT OR2C3 OR2L3 OR2W5P ZNF670 OR6F1 OR11L1 OR2M1P OR2L1P TRIM58 ZNF669 OR2L8 OR2T8 OR2G2 ZNF496 OR2L13 NLRP3 ZNF124 ZNF695 OR2AK2 OR2L2 OR9H1 OR2G3 OR14K1 OR2AJ1 GCSAML GCSAML-AS1 OR2L5
          nsv5310 Chr.1:247657184 - 247714043 on Build GRCh38 Deletion OR13G1 OR6F1
          nsv4066600 Chr.1:247670822 - 247920247 on Build GRCh38 Deletion OR11L1 OR1C1 TRIM58 OR9H1 OR13G1 OR14A2 OR14K1 OR6F1 OR14A16 OR2W3
          nsv4519110 Chr.1:247118697 - 247715698 on Build GRCh38 Duplication OR2B11 MIR3916 VN1R5 OR14L1 OR13G1 OR2G2 ZNF496 NLRP3 ZNF124 LOC102724446 ZNF496-DT OR2G3 OR2C3 OR2W5P OR6F1 GCSAML GCSAML-AS1
          nsv549598 Chr.1:247698214 - 247723700 on Build GRCh38 Loss OR14A2 OR6F1
          nsv6330035 Chr.1:247395130 - 247753305 on Build GRCh38 Duplication OR2B11 OR14L1 OR13G1 OR14A2 OR2G2 NLRP3 LOC102724446 OR2G3 OR2C3 OR2W5P OR14K1 OR6F1 GCSAML GCSAML-AS1

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline