Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01898369_cn
          See other IKBKE CNV Assays ›
          Gene Symbol
          IKBKE
          Assay Reference Genome
          Location

          Chr.1:206492763 on build GRCh38
          Cytoband
          Assay ID Hs01898369_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          9641

          Gene Name:

          inhibitor of nuclear factor kappa B kinase subunit epsilon

          Gene Aliases:

          IKK-E, IKK-i, IKKE, IKKI

          Location:

          Chr.1:206470476-206496889 on Build GRCh38

          Assay Gene Location:

          Within Intron 18
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          IKBKE NM_001193321.2 NP_001180250.1
          NM_001193322.2 NP_001180251.1
          NM_014002.4 NP_054721.1
          XM_005273356.3 XP_005273413.1
          AB016590.1 BAA85155.1
          AF241789.1 AAF45307.1
          AK093798.1
          BC105923.1 AAI05924.1
          BC105924.1 AAI05925.1
          BC107812.1 AAI07813.1
          D63485.1 BAA09772.3

          Target Gene Details

          Entrez Gene ID:

          574431

          Gene Name:

          IKBKE antisense RNA 1

          Gene Aliases:

          C1orf147

          Location:

          Chr.1:206491116-206497728 on Build GRCh38

          Assay Gene Location:

          Within Exon 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          IKBKE-AS1 NR_172918.1 2 449
          AK057159.1 2 449 BAB71372.1

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic Intronic Non-exonic non-DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of innate immune response
          response to stress
          immune response
          cellular response to DNA damage stimulus
          intrinsic apoptotic signaling pathway in response to DNA damage
          gene expression
          positive regulation of lipid storage
          positive regulation of type I interferon production
          response to cytokine
          response to type I interferon
          response to interferon-beta
          intracellular signal transduction
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of protein complex assembly
          mRNA stabilization
          defense response to virus
          type I interferon signaling pathway
          positive regulation of type I interferon-mediated signaling pathway
          interleukin-17-mediated signaling pathway
          cellular response to virus
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          IkappaB kinase activity
          kinase activity
          transferase activity
          protein phosphatase binding
          ubiquitin protein ligase binding
          K48-linked polyubiquitin binding
          identical protein binding
          K63-linked polyubiquitin binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline