Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02879866_cn
          See other LZTR1 CNV Assays ›
          Gene Symbol
          LZTR1
          Assay Reference Genome
          Location

          Chr.22:20995824 on build GRCh38
          Cytoband
          22q11.21
          Assay ID Hs02879866_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          8216

          Gene Name:

          leucine zipper like transcription regulator 1

          Gene Aliases:

          BTBD29, LZTR-1, NS10, SWNTS2

          Location:

          Chr.22:20982269-20999037 on Build GRCh38

          Assay Gene Location:

          Within Exon 17
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          LZTR1 NM_006767.3 17 2124 NP_006758.2
          AK296347.1 16 2005
          AK313502.1 12 1526
          AL833698.1 1 736
          BC026214.1 17 2026 AAH26214.2
          CT841521.1 17 2040 CAJ86451.1
          D38496.1 17 2019 BAA07508.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3893441 Chr.22:20733858 - 21135629 on Build GRCh38 Gain+Loss BCRP2 THAP7 PI4KA AIFM3 LRRC74B P2RX6P MIR649 P2RX6 CRKL LOC101928891 THAP7-AS1 SERPIND1 TUBA3FP LZTR1 SLC7A4 SNAP29
          nsv1065617 Chr.22:20705383 - 21111546 on Build GRCh38 Gain BCRP2 THAP7 PI4KA AIFM3 LRRC74B P2RX6P MIR649 P2RX6 CRKL LOC101928891 THAP7-AS1 SERPIND1 TUBA3FP LZTR1 SLC7A4 SNAP29
          dgv7994n54 Chr.22:20559470 - 21111491 on Build GRCh38 Gain SMPD4P1 BCRP2 THAP7 PI4KA AIFM3 TMEM191A LRRC74B P2RX6P MIR649 POM121L4P P2RX6 CRKL LOC101928891 THAP7-AS1 SERPIND1 TUBA3FP LZTR1 SLC7A4 SNAP29 MED15
          nsv834142 Chr.22:20979844 - 21170235 on Build GRCh38 Gain P2RX6 BCRP2 THAP7 THAP7-AS1 AIFM3 TUBA3FP LZTR1 SLC7A4 FAM230B LRRC74B P2RX6P MIR649
          nsv471185 Chr.22:20977267 - 21033678 on Build GRCh38 Loss P2RX6 THAP7 THAP7-AS1 AIFM3 TUBA3FP LZTR1 SLC7A4
          nsv834141 Chr.22:20859455 - 21051952 on Build GRCh38 Loss THAP7 AIFM3 LRRC74B P2RX6P MIR649 P2RX6 CRKL LOC101928891 THAP7-AS1 TUBA3FP LZTR1 SLC7A4 SNAP29
          nsv828961 Chr.22:20980653 - 21023359 on Build GRCh38 Gain P2RX6 THAP7 THAP7-AS1 AIFM3 TUBA3FP LZTR1
          esv2763693 Chr.22:20581182 - 21111546 on Build GRCh38 Gain SMPD4P1 BCRP2 THAP7 PI4KA AIFM3 TMEM191A LRRC74B P2RX6P MIR649 POM121L4P P2RX6 CRKL LOC101928891 THAP7-AS1 SERPIND1 TUBA3FP LZTR1 SLC7A4 SNAP29 MED15
          esv3647304 Chr.22:20902577 - 21081487 on Build GRCh38 Loss P2RX6 CRKL LOC101928891 THAP7 THAP7-AS1 AIFM3 TUBA3FP LZTR1 SLC7A4 LRRC74B P2RX6P MIR649
          nsv588306 Chr.22:20963144 - 21111491 on Build GRCh38 Gain P2RX6 LOC101928891 BCRP2 THAP7 THAP7-AS1 AIFM3 TUBA3FP LZTR1 SLC7A4 LRRC74B P2RX6P MIR649
          nsv588305 Chr.22:20721841 - 21108064 on Build GRCh38 Loss BCRP2 THAP7 PI4KA AIFM3 LRRC74B P2RX6P MIR649 P2RX6 CRKL LOC101928891 THAP7-AS1 SERPIND1 TUBA3FP LZTR1 SLC7A4 SNAP29
          esv2678175 Chr.22:20691629 - 21284480 on Build GRCh38 Deletion BCRP2 THAP7 PI4KA AIFM3 TMEM191A FAM230B LRRC74B P2RX6P MIR649 POM121L4P P2RX6 CRKL LOC101928891 THAP7-AS1 SERPIND1 TUBA3FP LZTR1 SLC7A4 GGT2 SNAP29 POM121L8P
          dgv7993n54 Chr.22:20346736 - 21111491 on Build GRCh38 Gain SMPD4P1 BCRP2 PI4KA KLHL22 P2RX6P P2RX6 CRKL LOC101928891 USP41 THAP7-AS1 SNAP29 THAP7 AIFM3 TMEM191A LRRC74B MIR649 POM121L4P ZNF74 SERPIND1 TUBA3FP LZTR1 LOC101927859 SLC7A4 SCARF2 MED15
          dgv4483n100 Chr.22:20362634 - 21212930 on Build GRCh38 Gain SMPD4P1 BCRP2 PI4KA KLHL22 FAM230B P2RX6P P2RX6 CRKL LOC101928891 USP41 THAP7-AS1 GGT2 SNAP29 THAP7 AIFM3 TMEM191A LRRC74B MIR649 POM121L4P ZNF74 SERPIND1 TUBA3FP LZTR1 SLC7A4 SCARF2 MED15
          nsv523162 Chr.22:20389246 - 21108064 on Build GRCh38 Loss SMPD4P1 BCRP2 THAP7 PI4KA KLHL22 AIFM3 TMEM191A LRRC74B P2RX6P MIR649 POM121L4P P2RX6 ZNF74 CRKL LOC101928891 USP41 THAP7-AS1 SERPIND1 TUBA3FP LZTR1 SLC7A4 SNAP29 SCARF2 MED15
          nsv828960 Chr.22:20953775 - 21057148 on Build GRCh38 Loss P2RX6 LOC101928891 THAP7 THAP7-AS1 AIFM3 TUBA3FP LZTR1 SLC7A4 LRRC74B P2RX6P MIR649

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          anatomical structure morphogenesis
          transcription factor activity, sequence-specific DNA binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0