Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03964847_cn
          See other SERPINF1 CNV Assays ›
          Gene Symbol
          SERPINF1
          Assay Reference Genome
          Location

          Chr.17:1766659 on build GRCh38
          Cytoband
          Assay ID Hs03964847_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5176

          Gene Name:

          serpin family F member 1

          Gene Aliases:

          EPC-1, OI12, OI6, PEDF, PIG35

          Location:

          Chr.17:190882-195947 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SERPINF1 NM_001329903.2 NP_001316832.1
          NM_001329904.2 NP_001316833.1
          NM_002615.7 NP_002606.3
          AA351026.1
          AB593011.1 BAJ83966.1
          AB593012.1 BAJ83967.1
          AB593013.1 BAJ83968.1
          AF400442.1 AAK92491.1
          AK315344.1
          AY513280.1 AAT08033.1
          BC000522.1 AAH00522.1
          BC013984.2 AAH13984.1
          CA405781.1
          HM005693.1 AEE61290.1
          M76979.1 AAA60058.1

          Target Gene Details

          Entrez Gene ID:

          11276

          Gene Name:

          synergin gamma

          Gene Aliases:

          AP1GBP1, SYNG

          Location:

          Chr.17:1753975-1848537 on Build GRCh38

          Assay Gene Location:

          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SYNRG

          Target Gene Details

          Entrez Gene ID:

          7473

          Gene Name:

          Wnt family member 3

          Gene Aliases:

          INT4, TETAMS

          Location:

          Chr.17:1281918-1338132 on Build GRCh38

          Assay Gene Location:

          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          WNT3

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv9490 Chr.17:1653428 - 1828993 on Build GRCh38 Loss SMYD4 MIR22HG SYNRG TLCD2 PRPF8 SERPINF1 MIR22 TADA2A WDR81 SERPINF2 WNT3 DUSP14 NSF
          nsv6509089 Chr.17:1765022 - 1773349 on Build GRCh38 Deletion SYNRG WNT3 SERPINF1
          nsv4237808 Chr.17:1582120 - 2528622 on Build GRCh38 Duplication OVCA2 MNT SMYD4 HNF1B SYNRG TSR1 SGSM2 ACACA SNORD91A SERPINF2 RILP METTL16 SMG6 RTN4RL1 MIR22HG MRPL45 DPH1 SNORD91B MIR212 DDX52 MIR22 YWHAEP7 SCARF1 TBC1D3E WNT3 LOC101927839 HIC1 RTN4RL1-AS1 RPA1 LOC101927864 GPR179 C17orf78 TBC1D3 MIR132 LOC284009 SERPINF1 ARHGAP23 WDR81 SOCS7 NPEPPSP1 SMG6-AS1 DUSP14 NSF SLC43A2 MIR378J LOC105371756 TLCD2 PRPF8 TADA2A SRR
          nsv1065585 Chr.17:1733183 - 1918769 on Build GRCh38 Gain MIR378J DDX52 WDR81 RPA1 SMYD4 SERPINF2 LOC105371756 SYNRG WNT3 DUSP14 NSF SERPINF1
          nsv1946 Chr.17:1717490 - 1781570 on Build GRCh38 Insertion TADA2A WDR81 SMYD4 SERPINF2 SYNRG WNT3 DUSP14 NSF SERPINF1
          nsv833335 Chr.17:1713955 - 1827844 on Build GRCh38 Gain+Loss MIR22 TADA2A WDR81 SMYD4 SERPINF2 MIR22HG SYNRG WNT3 DUSP14 NSF SERPINF1
          nsv6511128 Chr.17:1727705 - 1921432 on Build GRCh38 Duplication MIR378J DDX52 WDR81 RPA1 SMYD4 SERPINF2 LOC105371756 SYNRG WNT3 DUSP14 NSF SERPINF1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          short-term memory
          response to acidic pH
          negative regulation of endothelial cell migration
          negative regulation of gene expression
          positive regulation of neuron projection development
          negative regulation of angiogenesis
          ovulation cycle
          response to arsenic-containing substance
          positive regulation of neurogenesis
          retina development in camera-type eye
          epithelial cell proliferation involved in prostate gland development
          negative regulation of epithelial cell proliferation involved in prostate gland development
          cellular response to cobalt ion
          cellular response to retinoic acid
          cellular response to glucose stimulus
          cellular response to dexamethasone stimulus
          response to peptide
          serine-type endopeptidase inhibitor activity
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline