Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04088231_cn
          See other GUCD1 CNV Assays ›
          Gene Symbol
          GUCD1
          Assay Reference Genome
          Location

          Chr.22:24545228 on build GRCh38
          Cytoband
          Assay ID Hs04088231_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          83606

          Gene Name:

          guanylyl cyclase domain containing 1

          Gene Aliases:

          C22orf13, LLN4

          Location:

          Chr.22:24540438-24555894 on Build GRCh38

          Assay Gene Location:

          Within Intron 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          GUCD1 NM_001284251.2 NP_001271180.1
          NM_001284252.2 NP_001271181.1
          NM_001284253.2 NP_001271182.1
          NM_001284254.2 NP_001271183.1
          NM_001284255.2 NP_001271184.1
          NM_001284256.2 NP_001271185.1
          NM_001284257.2 NP_001271186.1
          NM_031444.4 NP_113632.2
          NR_104286.2
          XM_005261761.3 XP_005261818.1
          AK054681.1 BAB70791.1
          AK098754.1
          AK294972.1 BAG58044.1
          AK296893.1 BAG59452.1
          BC002924.2 AAH02924.2
          BC053529.1 AAH53529.1
          BC070109.1 AAH70109.1
          BQ893876.1
          BU153022.1
          BX346077.2
          DA033557.1
          HY059446.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv8039n54 Chr.22:23323169 - 24601506 on Build GRCh38 Gain DRICH1 UPB1 CHCHD10 FAM230I VPREB3 SPECC1L-ADORA2A GSTT2B LOC100652871 MMP11 GSTT4 GGT5 C22orf15 SUSD2 GSTT2 SPECC1L GGT1 SMARCB1 POM121L9P PCAT14 IGLL1 CES5AP1 MIF SNRPD3 RGL4 LINC01659 SLC2A11 ZNF70 LRRC75B MIF-AS1 ADORA2A-AS1 LOC124900476 GUCD1 CABIN1 ZDHHC8BP DDTL GUSBP11 ADORA2A DDT DERL3 LINC02556
          esv2724068 Chr.22:24288741 - 24669870 on Build GRCh38 Deletion SNRPD3 UPB1 ADORA2A LRRC75B SPECC1L-ADORA2A SPECC1L GGT1 ADORA2A-AS1 GUCD1 POM121L10P BCRP3
          nsv1055830 Chr.22:24005071 - 24643213 on Build GRCh38 Gain SNRPD3 UPB1 LRRC75B SPECC1L-ADORA2A ADORA2A-AS1 GUCD1 CABIN1 ADORA2A GSTT4 GGT5 SUSD2 SPECC1L GGT1 POM121L9P BCRP3
          nsv6060070 Chr.22:24288952 - 24667424 on Build GRCh38 Deletion SNRPD3 UPB1 ADORA2A LRRC75B SPECC1L-ADORA2A SPECC1L GGT1 ADORA2A-AS1 GUCD1 POM121L10P BCRP3
          nsv588744 Chr.22:24285587 - 24617379 on Build GRCh38 Gain SNRPD3 UPB1 ADORA2A LRRC75B SPECC1L-ADORA2A SPECC1L GGT1 ADORA2A-AS1 GUCD1
          nsv9884 Chr.22:24415625 - 24662249 on Build GRCh38 Gain+Loss SNRPD3 UPB1 ADORA2A LRRC75B SPECC1L-ADORA2A SPECC1L GGT1 ADORA2A-AS1 GUCD1 POM121L10P BCRP3
          nsv4536037 Chr.22:23625812 - 24669033 on Build GRCh38 Duplication DRICH1 UPB1 CHCHD10 VPREB3 SPECC1L-ADORA2A GSTT2B LOC100652871 MMP11 GSTT4 GGT5 C22orf15 SUSD2 GSTT2 SPECC1L GGT1 SMARCB1 POM121L9P POM121L10P MIF SNRPD3 RGL4 SLC2A11 ZNF70 LRRC75B MIF-AS1 ADORA2A-AS1 GUCD1 CABIN1 DDTL GUSBP11 ADORA2A DDT DERL3 BCRP3
          nsv1057819 Chr.22:24290518 - 24624081 on Build GRCh38 Gain SNRPD3 UPB1 ADORA2A LRRC75B SPECC1L-ADORA2A SPECC1L GGT1 ADORA2A-AS1 GUCD1
          esv2724041 Chr.22:23315914 - 24669376 on Build GRCh38 Deletion DRICH1 UPB1 CHCHD10 FAM230I VPREB3 SPECC1L-ADORA2A GSTT2B LOC100652871 MMP11 GSTT4 GGT5 C22orf15 SUSD2 GSTT2 SPECC1L GGT1 SMARCB1 POM121L9P PCAT14 POM121L10P IGLL1 CES5AP1 MIF SNRPD3 RGL4 LINC01659 SLC2A11 ZNF70 LRRC75B MIF-AS1 ADORA2A-AS1 LOC124900476 GUCD1 CABIN1 ZDHHC8BP DDTL GUSBP11 ADORA2A DDT BCR DERL3 LINC02556 BCRP3
          nsv526567 Chr.22:23348138 - 24600663 on Build GRCh38 Gain DRICH1 UPB1 CHCHD10 FAM230I VPREB3 SPECC1L-ADORA2A GSTT2B LOC100652871 MMP11 GSTT4 GGT5 C22orf15 SUSD2 GSTT2 SPECC1L GGT1 SMARCB1 POM121L9P PCAT14 IGLL1 CES5AP1 MIF SNRPD3 RGL4 LINC01659 SLC2A11 ZNF70 LRRC75B MIF-AS1 ADORA2A-AS1 LOC124900476 GUCD1 CABIN1 ZDHHC8BP DDTL GUSBP11 ADORA2A DDT DERL3
          nsv4288212 Chr.22:24545032 - 24550032 on Build GRCh38 Duplication GUCD1
          esv3893463 Chr.22:24009046 - 24601506 on Build GRCh38 Gain SNRPD3 UPB1 LRRC75B SPECC1L-ADORA2A ADORA2A-AS1 GUCD1 CABIN1 ADORA2A GGT5 SUSD2 SPECC1L GGT1 POM121L9P
          nsv6539653 Chr.22:22653980 - 24635071 on Build GRCh38 Duplication DRICH1 IGLV3-25 CHCHD10 SPECC1L-ADORA2A MIR5571 GSTT2B MMP11 GSTT4 GGT5 C22orf15 SUSD2 SPECC1L GGT1 SMARCB1 CES5AP1 RGL4 ADORA2A-AS1 LOC124900476 CABIN1 FBXW4P1 RAB36 GNAZ IGLV4-3 GUSBP11 ADORA2A BCR DERL3 IGLC1 BCRP3 UPB1 FAM230I VPREB3 IGLV2-5 LOC100652871 RSPH14 IGLJ3 GSTT2 MIR650 POM121L9P PCAT14 IGLL1 MIF SNRPD3 LINC01659 SLC2A11 ZNF70 LRRC75B MIF-AS1 IGLV3-19 GUCD1 ZDHHC8BP IGLL5 DDTL DDT LINC02556 IGLV3-21
          dgv4528n100 Chr.22:23322244 - 24682765 on Build GRCh38 Gain DRICH1 UPB1 CHCHD10 FAM230I VPREB3 SPECC1L-ADORA2A GSTT2B LOC100652871 MMP11 GSTT4 GGT5 C22orf15 SUSD2 GSTT2 SPECC1L GGT1 SMARCB1 POM121L9P PCAT14 POM121L10P IGLL1 CES5AP1 MIF SNRPD3 RGL4 LINC01659 SLC2A11 ZNF70 LRRC75B MIF-AS1 ADORA2A-AS1 LOC124900476 GUCD1 CABIN1 ZDHHC8BP DDTL GUSBP11 ADORA2A DDT DERL3 LINC02556 BCRP3
          dgv4649n223 Chr.22:23314752 - 24675047 on Build GRCh38 Duplication DRICH1 UPB1 CHCHD10 FAM230I VPREB3 SPECC1L-ADORA2A GSTT2B LOC100652871 MMP11 GSTT4 GGT5 C22orf15 SUSD2 GSTT2 SPECC1L GGT1 SMARCB1 POM121L9P PCAT14 POM121L10P IGLL1 CES5AP1 MIF SNRPD3 RGL4 LINC01659 SLC2A11 ZNF70 LRRC75B MIF-AS1 ADORA2A-AS1 LOC124900476 GUCD1 CABIN1 ZDHHC8BP DDTL GUSBP11 ADORA2A DDT BCR DERL3 LINC02556 BCRP3
          esv3575443 Chr.22:23348201 - 24670505 on Build GRCh38 Gain DRICH1 UPB1 CHCHD10 FAM230I VPREB3 SPECC1L-ADORA2A GSTT2B LOC100652871 MMP11 GSTT4 GGT5 C22orf15 SUSD2 GSTT2 SPECC1L GGT1 SMARCB1 POM121L9P PCAT14 POM121L10P IGLL1 CES5AP1 MIF SNRPD3 RGL4 LINC01659 SLC2A11 ZNF70 LRRC75B MIF-AS1 ADORA2A-AS1 LOC124900476 GUCD1 CABIN1 ZDHHC8BP DDTL GUSBP11 ADORA2A DDT DERL3 BCRP3
          dgv1387n209 Chr.22:22621614 - 24637144 on Build GRCh38 Deletion DRICH1 IGLV3-25 CHCHD10 SPECC1L-ADORA2A MIR5571 GSTT2B MMP11 GSTT4 GGT5 C22orf15 SUSD2 SPECC1L GGT1 SMARCB1 GGTLC2 CES5AP1 RGL4 ADORA2A-AS1 LOC124900476 CABIN1 FBXW4P1 RAB36 GNAZ IGLV4-3 GUSBP11 ADORA2A BCR DERL3 IGLC1 BCRP3 UPB1 POM121L1P FAM230I VPREB3 IGLV2-5 LOC100652871 RSPH14 IGLJ3 GSTT2 MIR650 POM121L9P PCAT14 IGLL1 MIF SNRPD3 LINC01659 SLC2A11 ZNF70 LRRC75B MIF-AS1 IGLV3-19 GUCD1 ZDHHC8BP IGLL5 DDTL DDT LINC02556 IGLV3-21

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs115115477] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          response to cAMP
          liver regeneration
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline