Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04197581_cn
          See other CFHR1 CNV Assays ›
          Gene Symbol
          CFHR1
          Assay Reference Genome
          Location

          Chr.1:196830173 on build GRCh38
          Cytoband
          Assay ID Hs04197581_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          3078

          Gene Name:

          complement factor H related 1

          Gene Aliases:

          CFHL, CFHL1, CFHL1P, CFHR1P, FHL-1, FHR-1, FHR1, H36, H36-1, H36-2, HFL1, HFL2

          Location:

          Chr.1:196819731-196832189 on Build GRCh38

          Assay Gene Location:

          Within Intron 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CFHR1 NM_001379306.1 NP_001366235.1
          NM_001379307.1 NP_001366236.1
          NM_001379308.1 NP_001366237.1
          NM_001379309.1 NP_001366238.1
          NM_001379310.1 NP_001366239.1
          NM_001379311.1 NP_001366240.1
          NM_001379312.1 NP_001366241.1
          NM_002113.3 NP_002104.2
          AK290830.1 BAF83519.1
          BC016755.1 AAH16755.1
          BC107771.1 AAI07772.1
          M65292.1 AAA35946.1
          M65293.1 AAA35947.1
          X56209.1 CAA39666.1
          X56210.1 CAA39667.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3891026 Chr.1:196743466 - 196851286 on Build GRCh38 Gain+Loss CFHR1 CFHR3 CFH
          dgv546n100 Chr.1:196819844 - 196847588 on Build GRCh38 Gain CFHR1
          dgv103n145 Chr.1:196827261 - 196949077 on Build GRCh38 Duplication CFHR2 CFHR1 CFHR4
          nsv548828 Chr.1:196812480 - 196844140 on Build GRCh38 Gain CFHR1
          dgv749n54 Chr.1:196808587 - 196835130 on Build GRCh38 Loss CFHR1
          dgv538n100 Chr.1:196787999 - 196843388 on Build GRCh38 Loss CFHR1 CFHR3
          nsv4068427 Chr.1:196742348 - 197084331 on Build GRCh38 Duplication ASPM F13B CFHR5 CFHR2 CFHR1 CFHR4 CFHR3 CFH
          dgv13e55 Chr.1:196764003 - 196932623 on Build GRCh38 Gain CFHR1 CFHR4 CFHR3
          nsv548801 Chr.1:196792630 - 196935320 on Build GRCh38 Loss CFHR1 CFHR4 CFHR3
          dgv508n100 Chr.1:196740618 - 196843388 on Build GRCh38 Loss CFHR1 CFHR3 CFH
          esv2421560 Chr.1:196753917 - 196846661 on Build GRCh38 Deletion CFHR1 CFHR3
          nsv3170002 Chr.1:196745743 - 196944667 on Build GRCh38 Deletion CFHR2 CFHR1 CFHR4 CFHR3 CFH
          nsv528460 Chr.1:196573437 - 197477572 on Build GRCh38 Gain ZBTB41 CRB1 MIR4735 ASPM F13B KCNT2 CFHR5 CFHR2 CFHR1 CFHR4 CFHR3 CFH
          esv33169 Chr.1:196769796 - 196853803 on Build GRCh38 Gain+Loss CFHR1 CFHR3
          nsv5445003 Chr.1:196755587 - 196846587 on Build GRCh38 Deletion CFHR1 CFHR3
          nsv3112209 Chr.1:196779326 - 196844127 on Build GRCh38 Deletion CFHR1 CFHR3
          nsv1010736 Chr.1:196801553 - 196832930 on Build GRCh38 Gain CFHR1
          nsv4053492 Chr.1:196821070 - 196836370 on Build GRCh38 Deletion CFHR1
          esv5113 Chr.1:196825198 - 196949317 on Build GRCh38 Loss CFHR2 CFHR1 CFHR4
          dgv550n100 Chr.1:196829423 - 196862182 on Build GRCh38 Gain CFHR1
          dgv43e201 Chr.1:196794400 - 196942973 on Build GRCh38 Deletion CFHR1 CFHR4 CFHR3
          nsv498700 Chr.1:196753075 - 196839376 on Build GRCh38 Loss CFHR1 CFHR3
          nsv3999 Chr.1:196748345 - 196951930 on Build GRCh38 Deletion CFHR2 CFHR1 CFHR4 CFHR3
          nsv548831 Chr.1:196826893 - 196852792 on Build GRCh38 Loss CFHR1
          esv26552 Chr.1:196741188 - 196950140 on Build GRCh38 Gain+Loss CFHR2 CFHR1 CFHR4 CFHR3 CFH
          nsv6328375 Chr.1:196703166 - 196869786 on Build GRCh38 Deletion CFHR1 CFHR3 CFH
          nsv548766 Chr.1:196748507 - 196932623 on Build GRCh38 Gain CFHR1 CFHR4 CFHR3
          esv3578358 Chr.1:196747283 - 196944121 on Build GRCh38 Loss CFHR2 CFHR1 CFHR4 CFHR3 CFH
          dgv88e199 Chr.1:196791346 - 196933087 on Build GRCh38 Deletion CFHR1 CFHR4 CFHR3
          nsv5982158 Chr.1:196817013 - 196934428 on Build GRCh38 Deletion CFHR1 CFHR4
          dgv733n54 Chr.1:196741937 - 196932623 on Build GRCh38 Loss CFHR1 CFHR4 CFHR3 CFH
          nsv826008 Chr.1:196739341 - 196859658 on Build GRCh38 Loss CFHR1 CFHR3 CFH
          dgv536n100 Chr.1:196787880 - 196843388 on Build GRCh38 Gain+Loss CFHR1 CFHR3
          nsv498701 Chr.1:196813583 - 196935541 on Build GRCh38 Loss CFHR1 CFHR4
          dgv3n31 Chr.1:196812967 - 196921215 on Build GRCh38 Gain CFHR1 CFHR4
          nsv428290 Chr.1:196621707 - 196992672 on Build GRCh38 Gain+Loss CFHR5 CFHR2 CFHR1 CFHR4 CFHR3 CFH
          nsv442555 Chr.1:196761905 - 196832942 on Build GRCh38 Loss CFHR1 CFHR3
          dgv532n100 Chr.1:196770830 - 196847588 on Build GRCh38 Gain CFHR1 CFHR3
          dgv547n100 Chr.1:196819859 - 196847588 on Build GRCh38 Loss CFHR1
          nsv437335 Chr.1:196824079 - 196833377 on Build GRCh38 Loss CFHR1
          dgv99n145 Chr.1:196758680 - 196851756 on Build GRCh38 Deletion CFHR1 CFHR3
          nsv1130635 Chr.1:196799084 - 196923949 on Build GRCh38 Deletion CFHR1 CFHR4
          nsv5879943 Chr.1:196761730 - 196846295 on Build GRCh38 Deletion CFHR1 CFHR3
          nsv819563 Chr.1:196747365 - 196843172 on Build GRCh38 Gain CFHR1 CFHR3 CFH
          dgv87e199 Chr.1:196753215 - 196853562 on Build GRCh38 Deletion CFHR1 CFHR3
          dgv747n54 Chr.1:196801995 - 196930960 on Build GRCh38 Loss CFHR1 CFHR4
          dgv750n54 Chr.1:196818614 - 196898103 on Build GRCh38 Loss CFHR1 CFHR4
          dgv539n100 Chr.1:196798270 - 196947657 on Build GRCh38 Gain CFHR2 CFHR1 CFHR4
          nsv820836 Chr.1:196747947 - 196852688 on Build GRCh38 Deletion CFHR1 CFHR3
          dgv734n54 Chr.1:196742052 - 196853900 on Build GRCh38 Loss CFHR1 CFHR3 CFH
          dgv492n223 Chr.1:196794612 - 196924144 on Build GRCh38 Duplication CFHR1 CFHR4 CFHR3
          nsv471614 Chr.1:196700381 - 196986100 on Build GRCh38 Gain+Loss CFHR5 CFHR2 CFHR1 CFHR4 CFHR3 CFH
          nsv1159841 Chr.1:196758720 - 196840786 on Build GRCh38 Deletion CFHR1 CFHR3
          nsv6139398 Chr.1:196756000 - 196840587 on Build GRCh38 Duplication CFHR1 CFHR3
          nsv471353 Chr.1:196819731 - 196832186 on Build GRCh38 Gain+Loss CFHR1
          esv1001500 Chr.1:196742223 - 196860102 on Build GRCh38 Deletion CFHR1 CFHR3 CFH
          nsv436214 Chr.1:196747584 - 196830482 on Build GRCh38 Deletion CFHR1 CFHR3
          dgv548n100 Chr.1:196820420 - 196832513 on Build GRCh38 Loss CFHR1
          nsv1159842 Chr.1:196780731 - 196940515 on Build GRCh38 Duplication CFHR1 CFHR4 CFHR3
          nsv469599 Chr.1:196827346 - 196986100 on Build GRCh38 Loss CFHR5 CFHR2 CFHR1 CFHR4
          esv3030456 Chr.1:196806870 - 196834870 on Build GRCh38 Deletion CFHR1
          dgv25n16 Chr.1:196808688 - 196941840 on Build GRCh38 Insertion CFHR1 CFHR4
          nsv1008827 Chr.1:196772225 - 196870033 on Build GRCh38 Loss CFHR1 CFHR3
          esv3544666 Chr.1:196752738 - 196852870 on Build GRCh38 Deletion CFHR1 CFHR3
          dgv503n100 Chr.1:196710325 - 196847588 on Build GRCh38 Gain CFHR1 CFHR3 CFH
          esv3584613 Chr.1:196740630 - 196832942 on Build GRCh38 Loss CFHR1 CFHR3 CFH
          dgv748n54 Chr.1:196805166 - 196964115 on Build GRCh38 Gain CFHR2 CFHR1 CFHR4
          dgv504n100 Chr.1:196729100 - 196884377 on Build GRCh38 Gain CFHR1 CFHR3 CFH
          dgv744n54 Chr.1:196787170 - 196835130 on Build GRCh38 Loss CFHR1 CFHR3
          dgv11e55 Chr.1:196762018 - 196932623 on Build GRCh38 Loss CFHR1 CFHR4 CFHR3
          nsv5573900 Chr.1:196830121 - 196830176 on Build GRCh38 Deletion CFHR1
          nsv1068779 Chr.1:196754369 - 196844570 on Build GRCh38 Deletion CFHR1 CFHR3
          esv2652513 Chr.1:196760586 - 196854711 on Build GRCh38 Loss CFHR1 CFHR3
          nsv5444857 Chr.1:196827000 - 196832000 on Build GRCh38 Duplication CFHR1
          nsv469810 Chr.1:196700380 - 196847241 on Build GRCh38 Loss CFHR1 CFHR3 CFH
          esv3588469 Chr.1:196759747 - 196839735 on Build GRCh38 Loss CFHR1 CFHR3
          nsv1118141 Chr.1:196799570 - 196833670 on Build GRCh38 Deletion CFHR1
          dgv505n100 Chr.1:196735867 - 196832930 on Build GRCh38 Gain CFHR1 CFHR3 CFH
          dgv42e201 Chr.1:196743570 - 196889169 on Build GRCh38 Deletion CFHR1 CFHR4 CFHR3 CFH
          dgv526n100 Chr.1:196769481 - 196855458 on Build GRCh38 Gain+Loss CFHR1 CFHR3
          dgv542n100 Chr.1:196814484 - 196847588 on Build GRCh38 Gain CFHR1
          dgv491n223 Chr.1:196788077 - 196926788 on Build GRCh38 Deletion CFHR1 CFHR4 CFHR3
          dgv746n54 Chr.1:196794012 - 196835130 on Build GRCh38 Loss CFHR1 CFHR3
          nsv946559 Chr.1:196820495 - 196855363 on Build GRCh38 Duplication CFHR1
          nsv514031 Chr.1:196769767 - 196832567 on Build GRCh38 Loss CFHR1 CFHR3
          dgv114n209 Chr.1:196797760 - 196942884 on Build GRCh38 Deletion CFHR1 CFHR4
          nsv3321483 Chr.1:196818001 - 196865100 on Build GRCh38 Duplication CFHR1
          esv3588478 Chr.1:196813225 - 196934743 on Build GRCh38 Loss CFHR1 CFHR4
          dgv98n145 Chr.1:196748999 - 196832286 on Build GRCh38 Deletion CFHR1 CFHR3
          dgv102n145 Chr.1:196787125 - 196926860 on Build GRCh38 Deletion CFHR1 CFHR4 CFHR3
          dgv741n54 Chr.1:196774797 - 196878339 on Build GRCh38 Loss CFHR1 CFHR3
          dgv490n223 Chr.1:196746801 - 196853068 on Build GRCh38 Deletion CFHR1 CFHR3 CFH
          nsv5570378 Chr.1:196757583 - 196842265 on Build GRCh38 Deletion CFHR1 CFHR3
          esv2630480 Chr.1:196757071 - 196842783 on Build GRCh38 Deletion CFHR1 CFHR3
          nsv3228719 Chr.1:196807116 - 196848329 on Build GRCh38 Insertion CFHR1
          nsv8691 Chr.1:196741634 - 196958768 on Build GRCh38 Gain+Loss CFHR2 CFHR1 CFHR4 CFHR3 CFH
          nsv3209410 Chr.1:196763652 - 196867679 on Build GRCh38 Deletion CFHR1 CFHR3
          nsv5445274 Chr.1:196814543 - 196935693 on Build GRCh38 Deletion CFHR1 CFHR4
          nsv6319124 Chr.1:196710401 - 197067700 on Build GRCh38 Duplication F13B CFHR5 CFHR2 CFHR1 CFHR4 CFHR3 CFH

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          complement activation
          negative regulation of protein binding
          cytolysis by host of symbiont cells
          complement component C3b binding
          protein binding
          identical protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline