Search
Search

Entrez Gene ID: | 10239 |
Gene Name: | adaptor related protein complex 3 subunit sigma 2 |
Gene Aliases: |
AP3S3, sigma3b |
Location: |
Chr.15:89830599-89893994 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP3S2 | NM_005829.5 | NP_005820.1 | ||
| NR_023361.2 | ||||
| NR_037582.2 | ||||
| AK222698.1 | BAD96418.1 | |||
| AK294712.1 | BAG57864.1 | |||
| AK312467.1 | BAG35374.1 | |||
| AL110108.1 | ||||
| BC002785.2 | AAH02785.1 | |||
| BC007773.2 | AAH07773.1 | |||
| BC010020.2 | AAH10020.1 | |||
| DA777624.1 | ||||
| DC340463.1 | ||||
| X99459.1 | CAA67824.1 |
Entrez Gene ID: | 100526783 |
Gene Name: | ARPIN-AP3S2 readthrough |
Gene Aliases: |
ARPIN, C15orf38, C15orf38-AP3S2 |
Location: |
Chr.15:89830599-89912952 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARPIN-AP3S2 | NM_001199058.2 | NP_001185987.1 | ||
| AK000495.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6496990 | Chr.15:89847380 - 89855969 on Build GRCh38 | Duplication |
|
| nsv6146172 | Chr.15:89845400 - 89854066 on Build GRCh38 | Deletion |
|
| esv3637203 | Chr.15:89845033 - 89856267 on Build GRCh38 | Loss |
|
Set Membership: |
|