Search
Search

| Assay Name: | aca-miR-31-5p |
|---|---|
| miRBase Accession Number: | MI0018852 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGCAAGAUGUUGGCAUAGCU |
| Species: | Anolis carolinensis, Capitella teleta, Lottia gigantea, Lytechinus variegatus, Patiria miniata, Python bivittatus, Strongylocentrotus purpuratus |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000064, mir-31 |
Mature miRNA Sequence: | AGGCAAGAUGUUGGCAUAGCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-miR-31-5p | MIMAT0021930 |
| Capitella teleta | cte-miR-31 | MIMAT0009515 |
| Lottia gigantea | lgi-miR-31 | MIMAT0009566 |
| Lytechinus variegatus | lva-miR-31-5p | MIMAT0032197 |
| Patiria miniata | pmi-miR-31-5p | MIMAT0032147 |
| Python bivittatus | pbv-miR-31-5p | MIMAT0038880 |
| View More Rows | ||
| Strongylocentrotus purpuratus | spu-miR-31 | MIMAT0009657 |