Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC14994
          Assay Name: aca-miR-31-5p
          miRBase Accession Number: MI0018852
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: AGGCAAGAUGUUGGCAUAGCU
          Species: Anolis carolinensis, Capitella teleta, Lottia gigantea, Lytechinus variegatus, Patiria miniata, Python bivittatus, Strongylocentrotus purpuratus
          Product Type: mirVana® miRNA mimic
          Assay ID MC14994

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000064, mir-31

          Mature miRNA Sequence:

          AGGCAAGAUGUUGGCAUAGCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Anolis carolinensis aca-miR-31-5p MIMAT0021930
          Capitella teleta cte-miR-31 MIMAT0009515
          Lottia gigantea lgi-miR-31 MIMAT0009566
          Lytechinus variegatus lva-miR-31-5p MIMAT0032197
          Patiria miniata pmi-miR-31-5p MIMAT0032147
          Python bivittatus pbv-miR-31-5p MIMAT0038880
          View More Rows
          Strongylocentrotus purpuratus spu-miR-31 MIMAT0009657

          Stem-loop Details



          Anolis carolinensis

          Stem-loop ID aca-mir-31
          Stem-loop Accession # MI0018852
          Stem-loop Sequence
          AUUCAGAGCUGGAGCGGAGGCAAGAUGUUGGCAUAGCUGAUGAGUUAAGAACCUGCUAUGCCAACAUAUUGCCCUCUUUCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021930
          miRBase ID: aca-miR-31-5p
          Mature miRNA Sequence: AGGCAAGAUGUUGGCAUAGCU
          Chromosome Location: NA

          Capitella teleta

          Stem-loop ID cte-mir-31
          Stem-loop Accession # MI0010059
          Stem-loop Sequence
          GCGAUCUCAGGGCACAGACCAGGCAAGAUGUUGGCAUAGCUGAUUUACACUCUGUGGUCAGCUCUGUCGCAUAUUGCCCCCUCUGGGCCCAGACUAACAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009515
          miRBase ID: cte-miR-31
          Mature miRNA Sequence: AGGCAAGAUGUUGGCAUAGCU
          Chromosome Location: NA

          Lottia gigantea

          Stem-loop ID lgi-mir-31
          Stem-loop Accession # MI0010109
          Stem-loop Sequence
          AUAAUGUUUGAGUCCAGACUAGGCAAGAUGUUGGCAUAGCUGAGUCAUAUAUGAACCAUCAGCUGUGUCUCAUAUUGCUAUCUUUGGGCUUGUUGUUGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009566
          miRBase ID: lgi-miR-31
          Mature miRNA Sequence: AGGCAAGAUGUUGGCAUAGCU
          Chromosome Location: NA

          Lytechinus variegatus

          Stem-loop ID lva-mir-31
          Stem-loop Accession # MI0025142
          Stem-loop Sequence
          UAUAAACCAGUGGAGAAAAGGCAAGAUGUUGGCAUAGCUGUGUGAUUUAAUACGAACCCAGCUGUGUCCUCAUAUUGCCAUUUAUUCACGUGGUUAUUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032197
          miRBase ID: lva-miR-31-5p
          Mature miRNA Sequence: AGGCAAGAUGUUGGCAUAGCU
          Chromosome Location: NA

          Patiria miniata

          Stem-loop ID pmi-mir-31-1
          Stem-loop Accession # MI0025093
          Stem-loop Sequence
          AGUGUAGAAAAGGCAAGAUGUUGGCAUAGCUGUUGUUGCCAAAAAACCAGCUGUACCUACAUAUUGCCAUUUGUCCACUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032147
          miRBase ID: pmi-miR-31-5p
          Mature miRNA Sequence: AGGCAAGAUGUUGGCAUAGCU
          Chromosome Location: NA

          Stem-loop ID pmi-mir-31-2
          Stem-loop Accession # MI0025094
          Stem-loop Sequence
          AGUGCCGAAGAGGCAAGAUGUUGGCAUAGCUGUGUCCAACGACAUAUUUUCAGCUGUGCCUACAUACUGCCAUUUUGUCACUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032147
          miRBase ID: pmi-miR-31-5p
          Mature miRNA Sequence: AGGCAAGAUGUUGGCAUAGCU
          Chromosome Location: NA

          Stem-loop ID pmi-mir-31-3
          Stem-loop Accession # MI0025095
          Stem-loop Sequence
          GUGGAGAAGGCAAGAUGUUGGCAUAGCUGUUUGAUUCAAAUUCUCAGCUGUGCUCACAUAUUGCCAUUUAUGCACUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032147
          miRBase ID: pmi-miR-31-5p
          Mature miRNA Sequence: AGGCAAGAUGUUGGCAUAGCU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-31
          Stem-loop Accession # MI0030189
          Stem-loop Sequence
          AUUGAGAGCUGGAGAGGAGGCAAGAUGUUGGCAUAGCUGAUGAACUGAGAACCUGCUAUGCCAACAUAUUGCCCUCUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038880
          miRBase ID: pbv-miR-31-5p
          Mature miRNA Sequence: AGGCAAGAUGUUGGCAUAGCU
          Chromosome Location: NA

          Strongylocentrotus purpuratus

          Stem-loop ID spu-mir-31
          Stem-loop Accession # MI0010193
          Stem-loop Sequence
          UACAAACCAGUGGAGAAAAGGCAAGAUGUUGGCAUAGCUGUGAUUUAAAUAUUAACCCAGCUGUGUCUUCAUACUGCCAUUUAUUCACUUGGUUGUUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009657
          miRBase ID: spu-miR-31
          Mature miRNA Sequence: AGGCAAGAUGUUGGCAUAGCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 006625_mat
          mirVana® miRNA inhibitor : MH14994


          miRBase Alias:

          Aeromonas caviae Ae398: aca-miR-31 (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline