Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002208
          Assay Name: hsa-miR-450b-3p
          miRBase Accession Number: MI0005531
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 134540185 - 134540262 [-] on Build GRCh38
          Mature miRNA Sequence: UUGGGAUCAUUUUGCAUCCAUA
          Species: Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002208
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000128, mir-450

          Mature miRNA Sequence:

          UUGGGAUCAUUUUGCAUCCAUA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-450b-3p MIMAT0004910
          Pan troglodytes ptr-miR-450b MIMAT0008152
          Pongo pygmaeus ppy-miR-450b-3p MIMAT0015895
          Rhesus monkey mml-miR-450b-3p MIMAT0006334

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-450b
          Stem-loop Accession # MI0005531
          Stem-loop Sequence
          GCAGAAUUAUUUUUGCAAUAUGUUCCUGAAUAUGUAAUAUAAGUGUAUUGGGAUCAUUUUGCAUCCAUAGUUUUGUAU
          Chromosome Location Chr. X - 134540185 - 134540262 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004910
          miRBase ID: hsa-miR-450b-3p
          Mature miRNA Sequence: UUGGGAUCAUUUUGCAUCCAUA
          Chromosome Location: Chr. X - 134540185 - 134540262 [-] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-450b
          Stem-loop Accession # MI0008674
          Stem-loop Sequence
          GCAGAAUUAUUUUUGCAAUAUGUUCCUGAAUAUGUAAUAUAAGUGUAUUGGGAUCAUUUUGCAUCCAUAGUUUUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008152
          miRBase ID: ptr-miR-450b
          Mature miRNA Sequence: UUGGGAUCAUUUUGCAUCCAUA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-450b
          Stem-loop Accession # MI0014951
          Stem-loop Sequence
          GCAGAAUUAUUUUUGCAAUAUGUUCCUGAAUAUGUAAUAUAAGUGUAUUGGGAUCAUUUUGCAUCCAUAGUUUUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015895
          miRBase ID: ppy-miR-450b-3p
          Mature miRNA Sequence: UUGGGAUCAUUUUGCAUCCAUA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-450b
          Stem-loop Accession # MI0007751
          Stem-loop Sequence
          GCAGAAUUAUUUUUGCAAUAUGUUCCUGAAUAUGUAGUAUAAGCGUAUUGGGAUCAUUUUGCAUCCAUAGUUUUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006334
          miRBase ID: mml-miR-450b-3p
          Mature miRNA Sequence: UUGGGAUCAUUUUGCAUCCAUA
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Human Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303731_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12749
          Ambion® Pre-miR™ miRNA Precursor : PM12749
          mirVana® miRNA inhibitor : MH12749
          mirVana® miRNA mimic : MC12749
          TaqMan™ Advanced miRNA Assay : 478913_mir
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          FGMIR : TaqManTM Human MicroRNA Array A
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline