Search
Search

| Assay Name: | hsa-miR-450b-3p |
|---|---|
| miRBase Accession Number: | MI0005531 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGGGAUCAUUUUGCAUCCAUA |
| Species: | Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000128, mir-450 |
Mature miRNA Sequence: | UUGGGAUCAUUUUGCAUCCAUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-450b-3p | MIMAT0004910 |
| Pan troglodytes | ptr-miR-450b | MIMAT0008152 |
| Pongo pygmaeus | ppy-miR-450b-3p | MIMAT0015895 |
| Rhesus monkey | mml-miR-450b-3p | MIMAT0006334 |