Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12749
          Assay Name: hsa-miR-450b-3p
          miRBase Accession Number: MI0005531
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 134540185 - 134540262 [-] on Build GRCh38
          Mature miRNA Sequence: UUGGGAUCAUUUUGCAUCCAUA
          Species: Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC12749

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000128, mir-450

          Mature miRNA Sequence:

          UUGGGAUCAUUUUGCAUCCAUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-450b-3p MIMAT0004910
          Pan troglodytes ptr-miR-450b MIMAT0008152
          Pongo pygmaeus ppy-miR-450b-3p MIMAT0015895
          Rhesus monkey mml-miR-450b-3p MIMAT0006334

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-450b
          Stem-loop Accession # MI0005531
          Stem-loop Sequence
          GCAGAAUUAUUUUUGCAAUAUGUUCCUGAAUAUGUAAUAUAAGUGUAUUGGGAUCAUUUUGCAUCCAUAGUUUUGUAU
          Chromosome Location Chr. X - 134540185 - 134540262 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004910
          miRBase ID: hsa-miR-450b-3p
          Mature miRNA Sequence: UUGGGAUCAUUUUGCAUCCAUA
          Chromosome Location: Chr. X - 134540185 - 134540262 [-] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-450b
          Stem-loop Accession # MI0008674
          Stem-loop Sequence
          GCAGAAUUAUUUUUGCAAUAUGUUCCUGAAUAUGUAAUAUAAGUGUAUUGGGAUCAUUUUGCAUCCAUAGUUUUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008152
          miRBase ID: ptr-miR-450b
          Mature miRNA Sequence: UUGGGAUCAUUUUGCAUCCAUA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-450b
          Stem-loop Accession # MI0014951
          Stem-loop Sequence
          GCAGAAUUAUUUUUGCAAUAUGUUCCUGAAUAUGUAAUAUAAGUGUAUUGGGAUCAUUUUGCAUCCAUAGUUUUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015895
          miRBase ID: ppy-miR-450b-3p
          Mature miRNA Sequence: UUGGGAUCAUUUUGCAUCCAUA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-450b
          Stem-loop Accession # MI0007751
          Stem-loop Sequence
          GCAGAAUUAUUUUUGCAAUAUGUUCCUGAAUAUGUAGUAUAAGCGUAUUGGGAUCAUUUUGCAUCCAUAGUUUUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006334
          miRBase ID: mml-miR-450b-3p
          Mature miRNA Sequence: UUGGGAUCAUUUUGCAUCCAUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303731_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12749
          Ambion® Pre-miR™ miRNA Precursor : PM12749
          TaqMan™ MicroRNA Assay : 002208
          mirVana® miRNA inhibitor : MH12749
          TaqMan™ Advanced miRNA Assay : 478913_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline