Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10042
          Assay Name: ath-miR398a-3p
          miRBase Accession Number: MI0001017
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 1040938 - 1041042 [+] on Build TAIR10.1
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Species: Arabidopsis, Aquilegia caerulea, Arabidopsis lyrata, Brassica oleracea, Citrus sinensis, Cucumis melo, Glycine max, Gossypium hirsutum, Gossypium raimondii, Malus domestica, Medicago truncatula, Populus trichocarpa, Rice, Ricinus communis, Theobroma cacao, Vitis vinifera
          Product Type: mirVana® miRNA mimic
          Assay ID MC10042

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000107, MIR398

          Mature miRNA Sequence:

          UGUGUUCUCAGGUCACCCCUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Arabidopsis ath-miR398a-3p MIMAT0000948
          Aquilegia caerulea aqc-miR398a MIMAT0012578
          Arabidopsis lyrata aly-miR398a-3p MIMAT0017566
          Brassica oleracea bol-miR398a-3p MIMAT0010174
          Citrus sinensis csi-miR398a-3p MIMAT0014079
          Cucumis melo cme-miR398b MIMAT0026149
          View More Rows
          Glycine max gma-miR398a MIMAT0001689
          Glycine max gma-miR398b-3p MIMAT0001690
          Gossypium hirsutum ghr-miR398 MIMAT0014337
          Gossypium raimondii gra-miR398 MIMAT0014328
          Malus domestica mdm-miR398a MIMAT0025998
          Medicago truncatula mtr-miR398a-3p MIMAT0011085
          Populus trichocarpa ptc-miR398a MIMAT0002041
          Rice osa-miR398a MIMAT0000982
          Ricinus communis rco-miR398a MIMAT0014194
          Theobroma cacao tcc-miR398b MIMAT0020434
          Vitis vinifera vvi-miR398a MIMAT0005727

          Stem-loop Details



          Arabidopsis

          Stem-loop ID ath-MIR398a
          Stem-loop Accession # MI0001017
          Stem-loop Sequence
          UGAAAUUUCAAAGGAGUGGCAUGUGAACACAUAUCCUAUGGUUUCUUCAAAUUUCCAUUGAAACCAUUGAGUUUUGUGUUCUCAGGUCACCCCUUUGAAUCUCCC
          Chromosome Location Chr. 2 - 1040938 - 1041042 [+] on Build TAIR10.1
          Mature MicroRNA miRBase Accession #: MIMAT0000948
          miRBase ID: ath-miR398a-3p
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: Chr. 2 - 1040938 - 1041042 [+] on Build TAIR10.1

          Aquilegia caerulea

          Stem-loop ID aqc-MIR398a
          Stem-loop Accession # MI0012096
          Stem-loop Sequence
          CCUGGAGGAGUGGCACUAAGAACACAGGUGGUUGUGCGAUUUCGAAUAUCAUUGCAUUUUGCAUUUGUGUUCUCAGGUCACCCCUUUGGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012578
          miRBase ID: aqc-miR398a
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA

          Arabidopsis lyrata

          Stem-loop ID aly-MIR398a
          Stem-loop Accession # MI0014585
          Stem-loop Sequence
          UAAUCAGUGGAAGUAAAGUAUGAAGAAAAAGAGAAGAACAACAGGAGGUGAAAUCUCAAAGGAGUGUCAUGAGAACACAGAUCCUAUGGUUUCUUUACAUUUUCAUUGAAACCAUUGAGUUUGUGUUCUCAGGUCACCCCUUUGAAUCUCCCCUGUUCCUUUAUUUUACCUUACCAAAAUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017566
          miRBase ID: aly-miR398a-3p
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA

          Brassica oleracea

          Stem-loop ID bol-MIR398a
          Stem-loop Accession # MI0010667
          Stem-loop Sequence
          UCUCAAAGGAGUGUCAUGAGAACACGGAUCCUCUUGUUUCUUUAAGAUUUCCACUGAAACCAUUGAGUAUUGUGUUCUCAGGUCACCCCUUUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010174
          miRBase ID: bol-miR398a-3p
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA

          Citrus sinensis

          Stem-loop ID csi-MIR398a
          Stem-loop Accession # MI0013305
          Stem-loop Sequence
          CAACAGAUGUGAAACCCCAGAGGAGUGAACCUGAGAACAGAGGGUGGCGUUGGCUUAAAUUUGAUUUGCAUUGCUGCUGCCUGCUCCUGCCAUACAUUAAAUCAAAAAGAUUGUAUGUGGCCAACGCACCUUGUGUUCUCAGGUCACCCCUUUGGGAAUUCAAUUGUCAAUCAGCACGUGUCAUCAAGUUACUCAAUUGCUCUAACAUUCGCUGCUUCUGAUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014079
          miRBase ID: csi-miR398a-3p
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA

          Cucumis melo

          Stem-loop ID cme-MIR398b
          Stem-loop Accession # MI0023253
          Stem-loop Sequence
          CCCAAAGGAGUGAACCCGAGAACACAAGAUUGCAUUUGAGAUUUGCCCAAUUGAUUGAUUUUUGUUUGUUGGGAGGAUUAAUUCAACAUAUUGCUCUUUGUGUUCUCAGGUCACCCCUUUGGAGACAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026149
          miRBase ID: cme-miR398b
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA

          Glycine max

          Stem-loop ID gma-MIR398a
          Stem-loop Accession # MI0001787
          Stem-loop Sequence
          CUCGGAGGAGUGAAUCUGAGAACACAAGGCUGGUUUGCACUGCUAUAUCAUCUAUUGGUAUAAGGUGAAUUUACUUUGUGUUCUCAGGUCACCCCUUUGAGCCAACCUGUUGACAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001689
          miRBase ID: gma-miR398a
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA

          Stem-loop ID gma-MIR398b
          Stem-loop Accession # MI0001788
          Stem-loop Sequence
          AGUCCAAAUGGUUUAUCUCAGAGGAGUGGAUCUGAGAACACAAGGCUGGUUUGCACUGCUAUAUUAUGAUCGAUUGGUAUAAGGUGAAUUUACUUUGUGUUCUCAGGUCACCCCUUUGAGCCAACCUGUUGACAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001690
          miRBase ID: gma-miR398b-3p
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA

          Gossypium hirsutum

          Stem-loop ID ghr-MIR398
          Stem-loop Accession # MI0013544
          Stem-loop Sequence
          GCCAAGAUGGUUUACCCCAGAGGGGUGCCACCUAAGAACACAGGUCAUCCCUUAACCAAAUAGCUUGUGUUCUCAGGUCACCCCUUUGGGGUACAUAUAUCUUCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014337
          miRBase ID: ghr-miR398
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA

          Gossypium raimondii

          Stem-loop ID gra-MIR398
          Stem-loop Accession # MI0013535
          Stem-loop Sequence
          GCCAAGAUGGUUUACCCCAGAGGGGUGCCACCUAAGAACACAGGUCAUCCCUUAACCAAAUAGCUUGUGUUCUCAGGUCACCCCUUUGGGGUACAUAUAUCUUCGCCAUUCCCUUUUGUUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014328
          miRBase ID: gra-miR398
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA

          Malus domestica

          Stem-loop ID mdm-MIR398a
          Stem-loop Accession # MI0023102
          Stem-loop Sequence
          GUUCAUAUAGCARAUGGUCACCCCAAAGGUGUGAAACUGAGAACACAAAUGGUGUUGYCYGCGACAAACAUUGUAAGCACUGCUUACGCAUUUUGUGUUCUCAGGUCACCCCUUUGGGGCACCUUGCUUACAUGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025998
          miRBase ID: mdm-miR398a
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA

          Medicago truncatula

          Stem-loop ID mtr-MIR398a
          Stem-loop Accession # MI0005599
          Stem-loop Sequence
          UCUCAGAGGAGUGACACUGAGAACACAAGAUUGAUUAAUCAUAUAAUGUAUUUGGUUGUUACUAGUUGAUUUUGUGUUCUCAGGUCACCCCUUUGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0011085
          miRBase ID: mtr-miR398a-3p
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA

          Populus trichocarpa

          Stem-loop ID ptc-MIR398a
          Stem-loop Accession # MI0002335
          Stem-loop Sequence
          GUACACCCCAGAGGAGUGGCUCCAGAGAACACAGGGGGUUGGUUUUCUAGCUGUAAGCUACAAGAUGGACAAAGCACUCUGUGUUCUCAGGUCACCCCUUUGGGGCACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002041
          miRBase ID: ptc-miR398a
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA

          Rice

          Stem-loop ID osa-MIR398a
          Stem-loop Accession # MI0001051
          Stem-loop Sequence
          GCUGAACCCAGAGGAGUGGUACUGAGAACACAGGUGCCAAUACAAUGUAUGGUGAGCUACUGUAUAAUGGAGUAAUUCUGUAACUGUGUUCUCAGGUCACCCCUUUGGGUUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0000982
          miRBase ID: osa-miR398a
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA

          Ricinus communis

          Stem-loop ID rco-MIR398a
          Stem-loop Accession # MI0013420
          Stem-loop Sequence
          CCAGAGGAGUGGCUCCCUGAGAACACACGUGCAUUGGCUCUCACCUUUUGUAGUUCCACUGCACCUGUGUUCUCAGGUCACCCCUUUGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014194
          miRBase ID: rco-miR398a
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA

          Theobroma cacao

          Stem-loop ID tcc-MIR398b
          Stem-loop Accession # MI0017518
          Stem-loop Sequence
          CAGCCAGAUGGUUUACCACGAAGGGGUGCCACCUAAGAACCCAGGCGGUUUGGCCUGAGCUAGGCCUUUGCCAAGCUGCUAUGAAAAGGAAGGCUGAAUGCUAAUGCAGCCAGCAGCAACCGGUUGUGUUCUCAGGUCACCCCUUCGCGGUACUUCUAUCUUCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0020434
          miRBase ID: tcc-miR398b
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA

          Vitis vinifera

          Stem-loop ID vvi-MIR398a
          Stem-loop Accession # MI0006572
          Stem-loop Sequence
          ACACCCCAAGGGAGUGGCACCUGAGAACACCGGUUGUGUUGGCUGCGCCAAGCUCUGCUAGCUAAUGCAUUUUGUGUUCUCAGGUCACCCCUUUGGGGCACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0005727
          miRBase ID: vvi-miR398a
          Mature miRNA Sequence: UGUGUUCUCAGGUCACCCCUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 000370
          mirVana® miRNA inhibitor : MH10042


          miRBase Alias:

          Arabidopsis thaliana: ath-miR398a (v20)
          Arabidopsis lyrata: aly-miR398a (v18)
          Brassica oleracea: bol-miR398a (v18)
          Citrus sinensis: csi-miR398 (v21)
          Glycine max: gma-miR398b (v21)
          Medicago truncatula: mtr-miR398a (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline