Search
Search

| Assay Name: | ath-miR398a-3p |
|---|---|
| miRBase Accession Number: | MI0001017 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUGUUCUCAGGUCACCCCUU |
| Species: | Arabidopsis, Aquilegia caerulea, Arabidopsis lyrata, Brassica oleracea, Citrus sinensis, Cucumis melo, Glycine max, Gossypium hirsutum, Gossypium raimondii, Malus domestica, Medicago truncatula, Populus trichocarpa, Rice, Ricinus communis, Theobroma cacao, Vitis vinifera |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000107, MIR398 |
Mature miRNA Sequence: | UGUGUUCUCAGGUCACCCCUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR398a-3p | MIMAT0000948 |
| Aquilegia caerulea | aqc-miR398a | MIMAT0012578 |
| Arabidopsis lyrata | aly-miR398a-3p | MIMAT0017566 |
| Brassica oleracea | bol-miR398a-3p | MIMAT0010174 |
| Citrus sinensis | csi-miR398a-3p | MIMAT0014079 |
| Cucumis melo | cme-miR398b | MIMAT0026149 |
| View More Rows | ||
| Glycine max | gma-miR398a | MIMAT0001689 |
| Glycine max | gma-miR398b-3p | MIMAT0001690 |
| Gossypium hirsutum | ghr-miR398 | MIMAT0014337 |
| Gossypium raimondii | gra-miR398 | MIMAT0014328 |
| Malus domestica | mdm-miR398a | MIMAT0025998 |
| Medicago truncatula | mtr-miR398a-3p | MIMAT0011085 |
| Populus trichocarpa | ptc-miR398a | MIMAT0002041 |
| Rice | osa-miR398a | MIMAT0000982 |
| Ricinus communis | rco-miR398a | MIMAT0014194 |
| Theobroma cacao | tcc-miR398b | MIMAT0020434 |
| Vitis vinifera | vvi-miR398a | MIMAT0005727 |