Search
Search

| Assay Name: | cgr-miR-28-5p |
|---|---|
| miRBase Accession Number: | MI0020475 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAGGAGCUCACAGUCUAUUGA |
| Species: | Cricetulus griseus, Goat, Pteropus alecto |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | AAGGAGCUCACAGUCUAUUGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Cricetulus griseus | cgr-miR-28-5p | MIMAT0023878 |
| Goat | chi-miR-28-5p | MIMAT0036109 |
| Pteropus alecto | pal-miR-28-5p | MIMAT0039890 |