Search
Search

| Assay Name: | lva-miR-29a-3p |
|---|---|
| miRBase Accession Number: | MI0025140 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAGCACCAGUUGAAAUCAGAGC |
| Species: | Lytechinus variegatus, Patiria miniata, Strongylocentrotus purpuratus |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | AAGCACCAGUUGAAAUCAGAGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Lytechinus variegatus | lva-miR-29a-3p | MIMAT0032195 |
| Patiria miniata | pmi-miR-29a-3p | MIMAT0032145 |
| Strongylocentrotus purpuratus | spu-miR-29a | MIMAT0009656 |