Search
Search

| Assay Name: | cfa-miR-653 |
|---|---|
| miRBase Accession Number: | MI0010423 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUGUUGAAACAAUCUCUAUUG |
| Species: | Dog |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000435, mir-653 |
Mature miRNA Sequence: | GUGUUGAAACAAUCUCUAUUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dog | cfa-miR-653 | MIMAT0009924 |