Search
Search

| Assay Name: | ppy-miR-615-3p |
|---|---|
| miRBase Accession Number: | MI0015100 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCGAGCCUGGGUCUCCGUCUU |
| Species: | Pongo pygmaeus |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000342, mir-615 |
Mature miRNA Sequence: | UCCGAGCCUGGGUCUCCGUCUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pongo pygmaeus | ppy-miR-615-3p | MIMAT0016057 |