Search
Search

| Assay Name: | ath-miR168a-3p |
|---|---|
| miRBase Accession Number: | MI0000210 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCCGCCUUGCAUCAACUGAAU |
| Species: | Arabidopsis, Arabidopsis lyrata, Brassica rapa, Camelina sativa, Citrus sinensis, Fragaria vesca, Medicago truncatula, Populus trichocarpa, Solanum lycopersicum |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000081, MIR168 |
Mature miRNA Sequence: | CCCGCCUUGCAUCAACUGAAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR168a-3p | MIMAT0031884 |
| Arabidopsis lyrata | aly-miR168a-3p | MIMAT0017482 |
| Brassica rapa | bra-miR168b-3p | MIMAT0035833 |
| Brassica rapa | bra-miR168c-3p | MIMAT0035835 |
| Camelina sativa | cas-miR168 | MIMAT0045309 |
| Citrus sinensis | csi-miR168-3p | MIMAT0048986 |
| View More Rows | ||
| Fragaria vesca | fve-miR168-3p | MIMAT0044511 |
| Medicago truncatula | mtr-miR168c-3p | MIMAT0022236 |
| Populus trichocarpa | ptc-miR168a-3p | MIMAT0022896 |
| Populus trichocarpa | ptc-miR168b-3p | MIMAT0022897 |
| Solanum lycopersicum | sly-miR168b-3p | MIMAT0031132 |