Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC19673
          Assay Name: ath-miR168a-3p
          miRBase Accession Number: MI0000210
          miRBase Version: v22.1
          Chromosome Location: Chr.4: 10578635 - 10578772 [+] on Build TAIR10.1
          Mature miRNA Sequence: CCCGCCUUGCAUCAACUGAAU
          Species: Arabidopsis, Arabidopsis lyrata, Brassica rapa, Camelina sativa, Citrus sinensis, Fragaria vesca, Medicago truncatula, Populus trichocarpa, Solanum lycopersicum
          Product Type: mirVana® miRNA mimic
          Assay ID MC19673

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000081, MIR168

          Mature miRNA Sequence:

          CCCGCCUUGCAUCAACUGAAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Arabidopsis ath-miR168a-3p MIMAT0031884
          Arabidopsis lyrata aly-miR168a-3p MIMAT0017482
          Brassica rapa bra-miR168b-3p MIMAT0035833
          Brassica rapa bra-miR168c-3p MIMAT0035835
          Camelina sativa cas-miR168 MIMAT0045309
          Citrus sinensis csi-miR168-3p MIMAT0048986
          View More Rows
          Fragaria vesca fve-miR168-3p MIMAT0044511
          Medicago truncatula mtr-miR168c-3p MIMAT0022236
          Populus trichocarpa ptc-miR168a-3p MIMAT0022896
          Populus trichocarpa ptc-miR168b-3p MIMAT0022897
          Solanum lycopersicum sly-miR168b-3p MIMAT0031132

          Stem-loop Details



          Arabidopsis

          Stem-loop ID ath-MIR168a
          Stem-loop Accession # MI0000210
          Stem-loop Sequence
          CACCAUCGGGCUCGGAUUCGCUUGGUGCAGGUCGGGAACCAAUUCGGCUGACACAGCCUCGUGACUUUUAAACCUUUAUUGGUUUGUGAGCAGGGAUUGGAUCCCGCCUUGCAUCAACUGAAUCGGAUCCUCGAGGUG
          Chromosome Location Chr. 4 - 10578635 - 10578772 [+] on Build TAIR10.1
          Mature MicroRNA miRBase Accession #: MIMAT0031884
          miRBase ID: ath-miR168a-3p
          Mature miRNA Sequence: CCCGCCUUGCAUCAACUGAAU
          Chromosome Location: Chr. 4 - 10578635 - 10578772 [+] on Build TAIR10.1

          Arabidopsis lyrata

          Stem-loop ID aly-MIR168a
          Stem-loop Accession # MI0014543
          Stem-loop Sequence
          GAGUCUCACCGUCGGGCUCGGAUUCGCUUGGUGCAGGUCGGGAACCAAUUCGGCUGACACAGCCUCGUGACUUUAAAACCUUUAUUGGUUUGUGAGCAGGGAUUGGAUCCCGCCUUGCAUCAACUGAAUCGGAUCCCCGAGGUGUAAGAAAACUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017482
          miRBase ID: aly-miR168a-3p
          Mature miRNA Sequence: CCCGCCUUGCAUCAACUGAAU
          Chromosome Location: NA

          Brassica rapa

          Stem-loop ID bra-MIR168b
          Stem-loop Accession # MI0030555
          Stem-loop Sequence
          CGGGCUCGGAUUCGCUUGGUGCAGGUCGGGACUAAUUCGCUGACACAGCCACGUGGAUUUUCCUUAUUGGUUUGUGAGCAGGGAUUGGAUCCCGCCUUGCAUCAACUGAAUCGGAGCCUCGAUGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035833
          miRBase ID: bra-miR168b-3p
          Mature miRNA Sequence: CCCGCCUUGCAUCAACUGAAU
          Chromosome Location: NA

          Stem-loop ID bra-MIR168c
          Stem-loop Accession # MI0030556
          Stem-loop Sequence
          GAAUCACCGUCGGGCUCGGAUUCGCUUGGUGCAGGUCGGGACACAAAUCACUGGUGCUUUUGCCUUAUUGGUUUGUGAGCAGGGAUUGGCUCCCGCCUUGCAUCAACUGAAUCGGAGCCUCCAAUGGUGAAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035835
          miRBase ID: bra-miR168c-3p
          Mature miRNA Sequence: CCCGCCUUGCAUCAACUGAAU
          Chromosome Location: NA

          Camelina sativa

          Stem-loop ID cas-MIR168
          Stem-loop Accession # MI0037196
          Stem-loop Sequence
          CUCGGAUUCGCUUGGUGCAGGUUGGGAACUGAUUGGCUGACACCGACUUUUGUUUUGUCGUCAUGGUUGGUUUGUCAGCUCCCGCCUUGCAUCAACUGAAUCGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0045309
          miRBase ID: cas-miR168
          Mature miRNA Sequence: CCCGCCUUGCAUCAACUGAAU
          Chromosome Location: NA

          Citrus sinensis

          Stem-loop ID csi-MIR168
          Stem-loop Accession # MI0039699
          Stem-loop Sequence
          GUUACCGGCGGUCUCUAAUUCGCUUGGUGCAGGUCGGGAACUGAUUGGCUAGUUUUUUUUUUGAAAUUUUUUGACAGCGAGGUGGCGUGUUGUAAUUGGUCGUAACGUUUAAAUUAAUGAUUGAAAAAAUGCGAAUUGGAUCCCGCCUUGCAUCAACUGAAUCGGAGACCGAGGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048986
          miRBase ID: csi-miR168-3p
          Mature miRNA Sequence: CCCGCCUUGCAUCAACUGAAU
          Chromosome Location: NA

          Fragaria vesca

          Stem-loop ID fve-MIR168
          Stem-loop Accession # MI0036379
          Stem-loop Sequence
          AGUAGUUACUGGCGGGUCUCUAAUUCGCUUGGUGCAGGUCGGGAACUGCUUCGGUUUUAGAGAGAGAGCCAGAUGGCUGCGGUUCCAGCGGAGCGGGUCCCGCCUUGCAUCAACUGAAUCGGAGACCGCGGUGAAAUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044511
          miRBase ID: fve-miR168-3p
          Mature miRNA Sequence: CCCGCCUUGCAUCAACUGAAU
          Chromosome Location: NA

          Medicago truncatula

          Stem-loop ID mtr-MIR168c
          Stem-loop Accession # MI0019093
          Stem-loop Sequence
          CUCACUUCGCGGUCUCUAAUUCGCUUGGUGCAGGUCGGGAACCACUACAUCCGCUGUUUUCGUAGAAACCGGCGGUGAAUUGGAUCCCGCCUUGCAUCAACUGAAUCGGAGGCCGCGGUGAAUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022236
          miRBase ID: mtr-miR168c-3p
          Mature miRNA Sequence: CCCGCCUUGCAUCAACUGAAU
          Chromosome Location: NA

          Populus trichocarpa

          Stem-loop ID ptc-MIR168a
          Stem-loop Accession # MI0002243
          Stem-loop Sequence
          GGUCUCUGAUUCGCUUGGUGCAGGUCGGGAACUGAUUCGGCGAUUUGAUUGCCAGAUGGCUCGACAUGACUGGUUGUUGUGGAAAAAGAAAAGGAAGGAAACAGGAAAAAAAACAAAGAAUAGCGAAUUGGAUCCCGCCUUGCAUCAACUGAAUCGGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022896
          miRBase ID: ptc-miR168a-3p
          Mature miRNA Sequence: CCCGCCUUGCAUCAACUGAAU
          Chromosome Location: NA

          Stem-loop ID ptc-MIR168b
          Stem-loop Accession # MI0002244
          Stem-loop Sequence
          GGUCUCUAAUUCGCUUGGUGCAGGUCGGGAACUGAUUCGGCGAUUUGAUUGCCAGAUGGCUAAACACGAUUGGCUGUGAGGCAAAUUAUAAAAAGAAAGAGAAUUGGAUCCCGCCUUGCAUCAACUGAAUCGGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022897
          miRBase ID: ptc-miR168b-3p
          Mature miRNA Sequence: CCCGCCUUGCAUCAACUGAAU
          Chromosome Location: NA

          Solanum lycopersicum

          Stem-loop ID sly-MIR168b
          Stem-loop Accession # MI0024353
          Stem-loop Sequence
          UUGCGGUGGUUUCUCAUUCGCUUGGUGCAGGUCGGGACCUACCUCGCCGGCAACAAUGGCGGUAGCUGACGGCGACGGCAGCGUACCGGUAAAAAAGAAUCUUAUUUUUACAUGUUGUUGCGUAGUUUGGGUCCCGCCUUGCAUCAACUGAAUAGGAGACUGCGGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031132
          miRBase ID: sly-miR168b-3p
          Mature miRNA Sequence: CCCGCCUUGCAUCAACUGAAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 465207_mat
          mirVana® miRNA inhibitor : MH19673


          miRBase Alias:

          Arabidopsis lyrata: aly-miR168a* (v18)
          Medicago truncatula: mtr-miR168c* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline