Search
Search

| Assay Name: | cca-miR6113 |
|---|---|
| miRBase Accession Number: | MI0021101 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCUGAAACUCAAGAACACGUUG |
| Species: | Cynara cardunculus |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UCUGAAACUCAAGAACACGUUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Cynara cardunculus | cca-miR6113 | MIMAT0024552 |