Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH37267
          Assay Name: mze-miR-150
          miRBase Accession Number: MI0034222
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: ACUCCCAAUCCUUGUACCAGUGUC
          Species: Metriaclima zebra, Neolamprologus brichardi, Oreochromis niloticus, Pundamilia nyererei
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH37267

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Mature miRNA Sequence:

          ACUCCCAAUCCUUGUACCAGUGUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Metriaclima zebra mze-miR-150 MIMAT0042416
          Neolamprologus brichardi nbr-miR-150 MIMAT0042598
          Oreochromis niloticus oni-miR-150 MIMAT0042787
          Pundamilia nyererei pny-miR-150 MIMAT0042987

          Stem-loop Details



          Metriaclima zebra

          Stem-loop ID mze-mir-150
          Stem-loop Accession # MI0034222
          Stem-loop Sequence
          ACUCCCAAUCCUUGUACCAGUGUCUUGACACCAGUGACGCUGGACGGGUUUUGGGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042416
          miRBase ID: mze-miR-150
          Mature miRNA Sequence: ACUCCCAAUCCUUGUACCAGUGUC
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-150
          Stem-loop Accession # MI0034478
          Stem-loop Sequence
          ACUCCCAAUCCUUGUACCAGUGUCUUGACACCAGUGACGCUGGACGGGUUUUGGGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042598
          miRBase ID: nbr-miR-150
          Mature miRNA Sequence: ACUCCCAAUCCUUGUACCAGUGUC
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-150
          Stem-loop Accession # MI0034733
          Stem-loop Sequence
          ACUCCCAAUCCUUGUACCAGUGUCUUGACACCAGUGACGCUGGACAGGUUUUGGGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042787
          miRBase ID: oni-miR-150
          Mature miRNA Sequence: ACUCCCAAUCCUUGUACCAGUGUC
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-150
          Stem-loop Accession # MI0034992
          Stem-loop Sequence
          ACUCCCAAUCCUUGUACCAGUGUCUUGACACCAGUGACGCUGGACGGGUUUUGGGGGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042987
          miRBase ID: pny-miR-150
          Mature miRNA Sequence: ACUCCCAAUCCUUGUACCAGUGUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          mirVana® miRNA inhibitor : MH37409
          mirVana® miRNA inhibitor : MH42130
          mirVana® miRNA inhibitor : MH42123
          mirVana® miRNA mimic : MC42123
          mirVana® miRNA mimic : MC42130
          mirVana® miRNA mimic : MC37267
          mirVana® miRNA mimic : MC37409

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0