Search
Search

| Assay Name: | mze-miR-150 |
|---|---|
| miRBase Accession Number: | MI0034222 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACUCCCAAUCCUUGUACCAGUGUC |
| Species: | Metriaclima zebra, Neolamprologus brichardi, Oreochromis niloticus, Pundamilia nyererei |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | ACUCCCAAUCCUUGUACCAGUGUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Metriaclima zebra | mze-miR-150 | MIMAT0042416 |
| Neolamprologus brichardi | nbr-miR-150 | MIMAT0042598 |
| Oreochromis niloticus | oni-miR-150 | MIMAT0042787 |
| Pundamilia nyererei | pny-miR-150 | MIMAT0042987 |