Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH37409
          Assay Name: mze-miR-150
          miRBase Accession Number: MI0034222
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: ACUCCCAAUCCUUGUACCAGUGUC
          Species: Metriaclima zebra, Neolamprologus brichardi, Oreochromis niloticus, Pundamilia nyererei
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH37409

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Mature miRNA Sequence:

          ACUCCCAAUCCUUGUACCAGUGUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Metriaclima zebra mze-miR-150 MIMAT0042416
          Neolamprologus brichardi nbr-miR-150 MIMAT0042598
          Oreochromis niloticus oni-miR-150 MIMAT0042787
          Pundamilia nyererei pny-miR-150 MIMAT0042987

          Stem-loop Details



          Metriaclima zebra

          Stem-loop ID mze-mir-150
          Stem-loop Accession # MI0034222
          Stem-loop Sequence
          ACUCCCAAUCCUUGUACCAGUGUCUUGACACCAGUGACGCUGGACGGGUUUUGGGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042416
          miRBase ID: mze-miR-150
          Mature miRNA Sequence: ACUCCCAAUCCUUGUACCAGUGUC
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-150
          Stem-loop Accession # MI0034478
          Stem-loop Sequence
          ACUCCCAAUCCUUGUACCAGUGUCUUGACACCAGUGACGCUGGACGGGUUUUGGGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042598
          miRBase ID: nbr-miR-150
          Mature miRNA Sequence: ACUCCCAAUCCUUGUACCAGUGUC
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-150
          Stem-loop Accession # MI0034733
          Stem-loop Sequence
          ACUCCCAAUCCUUGUACCAGUGUCUUGACACCAGUGACGCUGGACAGGUUUUGGGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042787
          miRBase ID: oni-miR-150
          Mature miRNA Sequence: ACUCCCAAUCCUUGUACCAGUGUC
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-150
          Stem-loop Accession # MI0034992
          Stem-loop Sequence
          ACUCCCAAUCCUUGUACCAGUGUCUUGACACCAGUGACGCUGGACGGGUUUUGGGGGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042987
          miRBase ID: pny-miR-150
          Mature miRNA Sequence: ACUCCCAAUCCUUGUACCAGUGUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          mirVana® miRNA inhibitor : MH42130
          mirVana® miRNA inhibitor : MH37267
          mirVana® miRNA inhibitor : MH42123
          mirVana® miRNA mimic : MC42123
          mirVana® miRNA mimic : MC42130
          mirVana® miRNA mimic : MC37267
          mirVana® miRNA mimic : MC37409

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline