Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CRH
Transcript:
NM_000756.3
Assay Name:
Mutation genome location:
chr.8 66177053-66177547 on build GRCh38
Assay ID
Hs00376382_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
peptide hormone
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
synaptic transmission, dopaminergic
glucocorticoid biosynthetic process
inflammatory response
signal transduction
chemical synaptic transmission
female pregnancy
parturition
learning or memory
positive regulation of cell proliferation
associative learning
hormone-mediated apoptotic signaling pathway
positive regulation of gene expression
negative regulation of gene expression
negative regulation of norepinephrine secretion
positive regulation of circadian sleep/wake cycle, wakefulness
positive regulation of cell death
regulation of serotonin secretion
diterpenoid metabolic process
hypothalamus development
lung development
adrenal gland development
positive regulation of cAMP biosynthetic process
negative regulation of epinephrine secretion
negative regulation of luteinizing hormone secretion
locomotory exploration behavior
positive regulation of insulin secretion involved in cellular response to glucose stimulus
response to immobilization stress
negative regulation of circadian sleep/wake cycle, REM sleep
response to drug
response to estrogen
response to ethanol
response to ether
negative regulation of blood pressure
response to pain
ion homeostasis
response to corticosterone
positive regulation of corticotropin secretion
positive regulation of cortisol secretion
long-term synaptic potentiation
positive regulation of digestive system process
negative regulation of cell death
negative regulation of glucagon secretion
cellular response to cocaine
cellular response to dexamethasone stimulus
positive regulation of calcium ion import
regulation of N-methyl-D-aspartate selective glutamate receptor activity
positive regulation of corticosterone secretion
positive regulation of behavioral fear response
receptor binding
hormone activity
neuropeptide hormone activity
protein binding
corticotropin-releasing hormone activity
corticotropin-releasing hormone receptor 1 binding
corticotropin-releasing hormone receptor 2 binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline