Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
MAPK14
Transcript:
NM_001315.2
Assay Name:
Mutation genome location:
chr.6 36059211-36059665 on build GRCh38
Assay ID
Hs00411838_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage checkpoint
activation of MAPK activity
cell morphogenesis
cartilage condensation
angiogenesis
placenta development
chondrocyte differentiation
glucose metabolic process
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
apoptotic process
movement of cell or subcellular component
chemotaxis
signal transduction
cell surface receptor signaling pathway
transmembrane receptor protein serine/threonine kinase signaling pathway
Ras protein signal transduction
skeletal muscle tissue development
response to glucose
positive regulation of gene expression
positive regulation of myotube differentiation
myoblast differentiation involved in skeletal muscle regeneration
peptidyl-serine phosphorylation
fatty acid oxidation
osteoclast differentiation
positive regulation of cyclase activity
lipopolysaccharide-mediated signaling pathway
response to muramyl dipeptide
intracellular signal transduction
cellular response to vascular endothelial growth factor stimulus
response to muscle stretch
p38MAPK cascade
positive regulation of protein import into nucleus
signal transduction in response to DNA damage
positive regulation of blood vessel endothelial cell migration
positive regulation of erythrocyte differentiation
positive regulation of myoblast differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of glucose import
protein autophosphorylation
vascular endothelial growth factor receptor signaling pathway
regulation of sequence-specific DNA binding transcription factor activity
striated muscle cell differentiation
positive regulation of muscle cell differentiation
positive regulation of cardiac muscle cell proliferation
3'-UTR-mediated mRNA stabilization
cellular response to lipopolysaccharide
cellular response to ionizing radiation
negative regulation of canonical Wnt signaling pathway
positive regulation of brown fat cell differentiation
stress-induced premature senescence
cellular response to virus
regulation of cytokine production involved in inflammatory response
positive regulation of myoblast fusion
regulation of signal transduction by p53 class mediator
positive regulation of reactive oxygen species metabolic process
positive regulation of interleukin-12 secretion
protein serine/threonine kinase activity
MAP kinase activity
MAP kinase kinase activity
protein binding
ATP binding
protein C-terminus binding
enzyme binding
protein phosphatase binding
NFAT protein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline