Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
HSP90AA1
Transcript:
NM_001017963.2
Assay Name:
Mutation genome location:
chr.14 102082006-102082599 on build GRCh38
Assay ID
Hs00461399_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
Hsp90 family chaperone
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
neuron migration
skeletal muscle contraction
mitochondrial transport
receptor-mediated endocytosis
response to unfolded protein
signal transduction
response to heat
response to cold
response to salt stress
positive regulation of lamellipodium assembly
cardiac muscle cell apoptotic process
peptidyl-tyrosine phosphorylation
regulation of protein ubiquitination
positive regulation of protein import into nucleus, translocation
Fc-gamma receptor signaling pathway involved in phagocytosis
ERBB2 signaling pathway
protein refolding
regulation of protein complex assembly
protein unfolding
response to estrogen
protein import into mitochondrial outer membrane
positive regulation of nitric oxide biosynthetic process
positive regulation of cell size
response to antibiotic
vascular endothelial growth factor receptor signaling pathway
protein stabilization
regulation of nitric-oxide synthase activity
chaperone-mediated protein complex assembly
positive regulation of cardiac muscle contraction
chaperone-mediated autophagy
regulation of cellular response to heat
nucleotide binding
UTP binding
CTP binding
mRNA binding
protein tyrosine kinase activity
protein binding
ATP binding
GTP binding
ATPase activity
sulfonylurea receptor binding
protein kinase binding
protein phosphatase binding
MHC class II protein complex binding
nitric-oxide synthase regulator activity
TPR domain binding
dATP binding
identical protein binding
protein homodimerization activity
histone deacetylase binding
ion channel binding
poly(A) RNA binding
GTPase binding
Rho GDP-dissociation inhibitor binding
unfolded protein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-7mgp5:80/100.66.78.208:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0