Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PTGER4
Transcript:
NM_000958.2
Assay Name:
Mutation genome location:
chr.5 40692317-40692812 on build GRCh38
Assay transcription exon location:
Within Exon 3
Assay ID
Hs00491707_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of peptide secretion
immune response
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
JNK cascade
female pregnancy
positive regulation of cell proliferation
response to mechanical stimulus
positive regulation of gene expression
negative regulation of hydrogen peroxide metabolic process
regulation of circadian sleep/wake cycle, wakefulness
positive regulation of smooth muscle cell migration
positive regulation of cAMP biosynthetic process
response to lipopolysaccharide
response to progesterone
positive regulation of interleukin-8 production
negative regulation of integrin activation
positive regulation of urine volume
positive regulation of renal sodium excretion
T-helper cell differentiation
negative regulation of circadian sleep/wake cycle, REM sleep
chemokinesis
response to drug
positive regulation of tyrosine phosphorylation of Stat3 protein
response to amino acid
positive regulation of osteoblast differentiation
positive regulation of bone resorption
positive regulation of cell adhesion
positive regulation of circadian sleep/wake cycle, non-REM sleep
negative regulation of cytokine secretion
negative regulation of interleukin-1 alpha secretion
positive regulation of cytokine secretion
negative regulation of inflammatory response
positive regulation of inflammatory response
regulation of stress fiber assembly
negative regulation of nitric-oxide synthase biosynthetic process
maternal process involved in parturition
bone development
positive regulation of mucus secretion
ERK1 and ERK2 cascade
cellular response to mechanical stimulus
cellular response to glucose stimulus
cellular response to interleukin-1
cellular response to prostaglandin E stimulus
positive regulation of wound healing
ductus arteriosus closure
positive regulation of hyaluronan biosynthetic process
response to salt
negative regulation of ductus arteriosus closure
negative regulation of small intestine smooth muscle contraction
positive regulation of calcitonin secretion
positive regulation of chemokinesis
positive regulation of matrix metallopeptidase secretion
negative regulation of tumor necrosis factor secretion
negative regulation of endothelin secretion
positive regulation of substance P secretion, neurotransmission
response to water-immersion restraint stress
positive regulation of antral ovarian follicle growth
positive regulation of neutrophil extravasation
negative regulation of eosinophil extravasation
positive regulation of interleukin-10 secretion
prostaglandin E receptor activity
protein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline