Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ATM
Transcript:
NM_000051.3
Assay Name:
Mutation genome location:
chr.11 108247009-108247167 on build GRCh38
Assay ID
Hs00532962_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Less than 200 bp
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
DNA double-strand break processing
DNA synthesis involved in DNA repair
strand displacement
response to hypoxia
somitogenesis
pre-B cell allelic exclusion
DNA replication
DNA repair
double-strand break repair via nonhomologous end joining
protein phosphorylation
cellular response to DNA damage stimulus
DNA damage induced protein phosphorylation
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
telomere maintenance via telomerase
cell cycle arrest
mitotic spindle assembly checkpoint
reciprocal meiotic recombination
signal transduction
brain development
heart development
determination of adult lifespan
intrinsic apoptotic signaling pathway in response to DNA damage
response to ionizing radiation
regulation of autophagy
histone phosphorylation
peptidyl-serine phosphorylation
negative regulation of B cell proliferation
positive regulation of telomere maintenance via telomerase
positive regulation of histone phosphorylation
V(D)J recombination
phosphatidylinositol-3-phosphate biosynthetic process
peptidyl-serine autophosphorylation
lipoprotein catabolic process
regulation of apoptotic process
positive regulation of apoptotic process
positive regulation of DNA damage response, signal transduction by p53 class mediator
positive regulation of neuron apoptotic process
meiotic telomere clustering
protein autophosphorylation
oocyte development
neuron apoptotic process
regulation of telomerase activity
histone mRNA catabolic process
cellular response to gamma radiation
cellular response to X-ray
cellular response to nitrosative stress
signal transduction involved in mitotic G2 DNA damage checkpoint
replicative senescence
establishment of RNA localization to telomere
establishment of macromolecular complex localization to telomere
regulation of cellular response to heat
regulation of signal transduction by p53 class mediator
negative regulation of TORC1 signaling
negative regulation of telomere capping
positive regulation of telomere maintenance via telomere lengthening
positive regulation of telomerase catalytic core complex assembly
DNA binding
protein serine/threonine kinase activity
DNA-dependent protein kinase activity
protein binding
ATP binding
1-phosphatidylinositol-3-kinase activity
protein complex binding
protein dimerization activity
protein N-terminus binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline