Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PCDHGA1
Transcript:
NM_032088.1
Assay Name:
Mutation genome location:
chr.5 141395156-141395381 on build GRCh38
Gene:
PCDHGA2
Transcript:
NM_032088.1
Assay Name:
Mutation genome location:
chr.5 141395156-141395381 on build GRCh38
Gene:
PCDHGA3
Transcript:
NM_032088.1
Assay Name:
Mutation genome location:
chr.5 141395156-141395381 on build GRCh38
Gene:
PCDHGA4
Transcript:
NM_032088.1
Assay Name:
Mutation genome location:
chr.5 141395156-141395381 on build GRCh38
Gene:
PCDHGA5
Transcript:
NM_032088.1
Assay Name:
Mutation genome location:
chr.5 141395156-141395381 on build GRCh38
Gene:
PCDHGA6
Transcript:
NM_032088.1
Assay Name:
Mutation genome location:
chr.5 141395156-141395381 on build GRCh38
Gene:
PCDHGA7
Transcript:
NM_032088.1
Assay Name:
Mutation genome location:
chr.5 141395156-141395381 on build GRCh38
Gene:
PCDHGA8
Transcript:
NM_032088.1
Assay Name:
Mutation genome location:
chr.5 141395156-141395381 on build GRCh38
Gene:
PCDHGB1
Transcript:
NM_032088.1
Assay Name:
Mutation genome location:
chr.5 141395156-141395381 on build GRCh38
Gene:
PCDHGB2
Transcript:
NM_032088.1
Assay Name:
Mutation genome location:
chr.5 141395156-141395381 on build GRCh38
Gene:
PCDHGB3
Transcript:
NM_032088.1
Assay Name:
Mutation genome location:
chr.5 141395156-141395381 on build GRCh38
Gene:
PCDHGB4
Transcript:
NM_032088.1
Assay Name:
Mutation genome location:
chr.5 141395156-141395381 on build GRCh38
Assay ID
Hs00592170_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
Process(es)
homophilic cell adhesion via plasma membrane adhesion molecules
endosome to lysosome transport
lipid tube assembly involved in organelle fusion
cell adhesion
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
calcium ion binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline