Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
SCARB1
Transcript:
ENST00000261693.0
Assay Name:
Mutation genome location:
chr.12 124811773-124812046 on build GRCh38
Assay ID
Hs00824468_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
endothelial cell proliferation
androgen biosynthetic process
cholesterol catabolic process
receptor-mediated endocytosis
positive regulation of endothelial cell migration
positive regulation of triglyceride biosynthetic process
positive regulation of cholesterol storage
regulation of phosphatidylcholine catabolic process
phospholipid transport
lipopolysaccharide transport
lipopolysaccharide-mediated signaling pathway
detection of lipopolysaccharide
cholesterol efflux
high-density lipoprotein particle remodeling
low-density lipoprotein particle clearance
high-density lipoprotein particle clearance
vitamin transmembrane transport
wound healing
lipoprotein metabolic process
cholesterol homeostasis
blood vessel endothelial cell migration
recognition of apoptotic cell
reverse cholesterol transport
adhesion of symbiont to host
viral entry into host cell
regulation of phagocytosis
intestinal absorption
positive regulation of nitric-oxide synthase activity
triglyceride homeostasis
cholesterol import
lipopolysaccharide binding
virus receptor activity
phosphatidylserine binding
lipopolysaccharide receptor activity
transporter activity
protein binding
1-phosphatidylinositol binding
high-density lipoprotein particle binding
low-density lipoprotein particle binding
apolipoprotein binding
apolipoprotein A-I binding
protein homodimerization activity
high-density lipoprotein particle receptor activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline