Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023464.2 | NP_075953.1 | 2 | 197 | ||
GenBank mRNA | AB041656.1 | - | 2 | 191 | ||
AK003356.2 | - | 2 | 198 | |||
AK088418.1 | - | 2 | 129 | |||
AK158412.1 | - | 314 | ||||
BC023033.1 | - | 2 | 160 | |||
BC037081.1 | - | 2 | 174 | 5098653 | ||
BY703613.1 | - | 2 | 198 |