Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF396 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322286.1 | NP_001309215.1 | 4 | 3237 | ||
NM_001322290.1 | NP_001309219.1 | 5 | 3474 | |||
NM_145756.2 | NP_665699.1 | 5 | 2367 | |||
GenBank mRNA | AK055775.1 | - | 5 | 2371 | ||
BC130288.1 | - | 5 | 2360 | 40146319 | ||
BC136314.1 | - | 5 | 2360 | 9020301 |